Skip to Content
Merck

Skip To

MX0480

Methanol

≥99.9% (GC), suitable for LC/MS, gas chromatography (GC), HPLC, OmniSolv®

Synonym(s):

Methyl alcohol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
CH3OH
CAS Number:
Molecular Weight:
32.04
UNSPSC Code:
41116121
EC Index Number:
200-659-6
MDL number:
Beilstein/REAXYS Number:
1098229
Assay:
≥99.9% (GC)
Technique(s):
HPLC: suitable, LC/MS: suitable, gas chromatography (GC): suitable
Bp:
64.7 °C (lit.)
Vapor pressure:
128 hPa ( 20 °C), 410 mmHg ( 50 °C), 97.68 mmHg ( 20 °C)
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Methanol, OmniSolv® HR-GC

vapor density

1.11 (vs air)

Quality Segment

vapor pressure

128 hPa ( 20 °C), 410 mmHg ( 50 °C), 97.68 mmHg ( 20 °C)

product line

OmniSolv®

assay

≥99.9% (GC)

form

liquid

autoignition temp.

725 °F

expl. lim.

36 %

technique(s)

HPLC: suitable, LC/MS: suitable, gas chromatography (GC): suitable

impurities

≤0.05% Water (H2O), ≤0.3 μeq/g Titrable acid, ≤1 ppm Formaldehyde (HCHO)

evapn. residue

≤1 ppm

color

clearAPHA: ≤5

refractive index

n20/D 1.329 (lit.)

bp

64.7 °C (lit.)

mp

−98 °C (lit.)

transition temp

flash point 9.7 °C

density

0.791 g/mL at 25 °C (lit.)

format

neat

shipped in

ambient

storage temp.

room temp

SMILES string

CO

InChI

1S/CH4O/c1-2/h2H,1H3

InChI key

OKKJLVBELUTLKV-UHFFFAOYSA-N

General description

Methanol, also known as Methyl alcohol, is a colorless organic solvent commonly used to dissolve substances with high and medium polarity.(3)

Application

Methanol is used as
  • A solvent to prepare the Mobile phase for LC-MS/MS analysis.
  • A disinfectant to clean laboratory space.
OmniSolv® HR-GC

Packaging

M-Bottle for Solvents
As a global leader in lab reagents, we are constantly looking for new ways to optimize the safety of our products. The newly developed 4L solvent bottle design features advanced sealing technology that eliminates leaks to make the handling of solvents safer and more convenient than ever before.
See all the new features here!
is packaged in 1L amber glass bottle
is packaged in 4L amber glass bottle

Preparation Note

Product filtered through a 0.2 μm filter

Analysis Note

AppearanceClear liquid, free from particulatesAssay (GC)99.9% minCap. ECD responsive substances as PCNB (peak)2ng/L maxCap. ECD responsive substances as PCNB (total)20ng/L maxCap. FID responsive substances as decane (peak)1µg/L maxCap. FID responsive substances as decane (total)10µg/L maxColor (APHA)5 maxEvaporation residue1ppm maxFiltered through 0.2 µm filterTo pass testFormaldehyde (HCHO)1ppm maxInfrared SpectrumConforms to standardTitrable acid0.3µeq/g maxWater (H2O)0.05% max
Evaporation Number: 1.9

Legal Information

OMNISOLV is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
MX04861.03726320390
assay

≥99.9% (GC)

assay

≥99.9% (GC)

assay

99.9% (GC)

assay

≥99.8%

technique(s)

HPLC: suitable, gas chromatography (GC): suitable, LC/MS: suitable

technique(s)

HPLC: suitable, LC/MS: suitable

technique(s)

HPLC: suitable, LC/MS: suitable, UHPLC: suitable

technique(s)

-

Quality Level

100

Quality Level

100

Quality Level

200

Quality Level

200

form

liquid

form

liquid

form

liquid

form

liquid

vapor pressure

128 hPa ( 20 °C), 97.68 mmHg ( 20 °C), 410 mmHg ( 50 °C)

vapor pressure

128 hPa ( 20 °C), 410 mmHg ( 50 °C), 97.68 mmHg ( 20 °C)

vapor pressure

128 hPa ( 20 °C), 410 mmHg ( 50 °C), 97.68 mmHg ( 20 °C)

vapor pressure

410 mmHg ( 50 °C), 97.68 mmHg ( 20 °C)

impurities

≤0.05% Water (H2O), ≤1 ppm Formaldehyde (HCHO), ≤0.3 μeq/g Titrable acid

impurities

≤0.05% Water (H2O), ≤0.2 μeq/g Titrable base, ≤0.3 μeq/g Titrable acid, ≤50 ppb p-nitrophenol (LC/MS), ≤50 ppb reserpine (LC/MS)

impurities

-

impurities

H2SO4, passes test (darkened), MnO4- reducers, passes test, ≤0.0002 meq/g Titr. base, ≤0.0003 meq/g Titr. acid, ≤0.001% acetaldehyde, ≤0.001% acetone, ≤0.001% formaldehyde, ≤0.1% water


Still not finding the right product?

Explore all of our products under Methanol


signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

target_organs

Eyes,Central nervous system

Storage Class

3 - Flammable liquids

wgk

WGK 2

flash_point_f

49.5 °F - closed cup

flash_point_c

9.7 °C - closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters