Skip to Content
Merck
Methanol low benzene, ACS reagent, ≥99.8%

Methanol

Methanol low benzene, ACS reagent, ≥99.8%
Synonym(s):
Methyl alcohol, Methanol
Linear Formula:
CH3OH
CAS Number:
Molecular Weight:
32.04
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Product Line
Purity
Agency
Format
Quality Segment
Application
Special Grade
Technique
Showing 1-20 of 56
Page 1 of 3
Compare
Product Number
Description
Pricing
Page 1 of 3