Skip to Content
Merck

Skip To

MX0475

Methanol

≥99.8% (GC), suitable for HPLC, meets USP/NF

Synonym(s):

Methyl alcohol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Linear Formula:
CH3OH
CAS Number:
Molecular Weight:
32.04
UNSPSC Code:
41116121
EC Index Number:
200-659-6
MDL number:
Beilstein/REAXYS Number:
1098229
Assay:
≥99.8% (GC)
Grade:
HPLC grade
Technique(s):
HPLC: suitable
Bp:
64.7 °C (lit.)
Vapor pressure:
128 hPa ( 20 °C), 410 mmHg ( 50 °C), 97.68 mmHg ( 20 °C)
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Methanol, HPLC, Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs

grade

HPLC grade

Quality Level

agency

meets USP/NF

vapor density

1.11 (vs air)

vapor pressure

128 hPa ( 20 °C), 410 mmHg ( 50 °C), 97.68 mmHg ( 20 °C)

assay

≥99.8% (GC)

form

liquid

autoignition temp.

725 °F

expl. lim.

36 %

technique(s)

HPLC: suitable

impurities

≤0.0002 meq/g Titrable base, ≤0.0003 meq/g Titrable acid, ≤0.001% Carbonyl (acetone,formaldehyde,acetaldehyde), ≤0.05% Water (H2O)

evapn. residue

≤2 ppm

color

APHA: ≤10

refractive index

n20/D 1.329 (lit.)

bp

64.7 °C (lit.)

mp

−98 °C (lit.)

transition temp

flash point 9.7 °C

density

0.791 g/mL at 25 °C (lit.)

UV cutoff

≤205 nm

UV absorption

λ: 205 nm Amax: ≤1.00, λ: 210 nm Amax: ≤0.80, λ: 220 nm Amax: ≤0.40, λ: 230 nm Amax: ≤0.20, λ: 240 nm Amax: ≤0.10, λ: 260 nm Amax: ≤0.04, λ: 280-400 nm Amax: ≤0.01

application(s)

PFAS testing

format

neat

shipped in

ambient

storage temp.

room temp

SMILES string

CO

InChI

1S/CH4O/c1-2/h2H,1H3

InChI key

OKKJLVBELUTLKV-UHFFFAOYSA-N

General description

Methanol is suitable for isocratic HPLC, meets ACS specifications for testing USP/NF testing monographs. It is extensively utilized in High-Performance Liquid Chromatography (HPLC) due to its excellent solvation properties and high miscibility with both water and organic solvents, thus making it an effective solvent for preparing mobile phases that enhance the separation and sensitivity of analyte detection.

Application

Methanol can be used as:
  • A solvent for the extraction of pesticides from soil samples.
  • An organic modifier in the quantification of esculin in pharmaceutical formulations via reverse-phase high-performance liquid chromatography (RP-HPLC).
HPLC, Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs
MX0475P-6 and MX0475P-1 are packaged in poly coated glass bottles
MX0475-NP10 is sold as 2 x 10 L

Packaging

M-Bottle for Solvents
As a global leader in lab reagents, we are constantly looking for new ways to optimize the safety of our products. The newly developed 4L solvent bottle design features advanced sealing technology that eliminates leaks to make the handling of solvents safer and more convenient than ever before.
See all the new features here!

Preparation Note

Product filtered through a 0.2 μm filter

Analysis Note

AppearanceClear liquid free from particulatesAssay (GC)99.8% minCarbonyl (acetone,formaldehyde,acetaldehyde)0.001% maxColor (APHA)10 maxEvaporation residue2ppm maxFiltered through 0.2 µm filterTo pass testGradient at 254 nm0.005AU maxIdentity (IR-spectrum)ConformsSolubility in waterTo pass testSubstances darkened by sulfuric acidTo pass testSubstances reducing permanganateTo pass testTitrable acid0.0003meq/g maxTitrable base0.0002meq/g maxUV Abs. at 205 nm1.00AU maxUV Abs. at 210 nm0.80AU maxUV Abs. at 220 nm0.40AU maxUV Abs. at 230 nm0.20AU maxUV Abs. at 240 nm0.10AU maxUV Abs. at 260 nm0.04AU maxUV Abs. at 280 - 400 nm0.01AU maxUV Cut-off205nm maxWater (H2O)0.05% max
Evaporation Number: 1.9

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
9006886463771.03726
assay

≥99.8% (GC)

assay

≥99.92%

assay

≥99.9%

assay

99.9% (GC)

technique(s)

HPLC: suitable

technique(s)

HPLC: suitable, LC/MS: suitable, UHPLC: suitable, mass spectrometry (MS): suitable

technique(s)

HPLC: suitable

technique(s)

HPLC: suitable, LC/MS: suitable, UHPLC: suitable

application(s)

PFAS testing

application(s)

PFAS testing
food and beverages

application(s)

PFAS testing

application(s)

PFAS testing

Quality Level

100, 300

Quality Level

100

Quality Level

100

Quality Level

200

grade

HPLC grade

grade

HPLC grade

grade

HPLC Plus

grade

-

form

liquid

form

liquid

form

liquid

form

liquid


Still not finding the right product?

Explore all of our products under Methanol


signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

target_organs

Eyes,Central nervous system

Storage Class

3 - Flammable liquids

flash_point_f

49.5 °F - closed cup

flash_point_c

9.7 °C - closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Development of a robust and reliable reverse-phase high-performance liquid chromatography (RP-HPLC) method using analytical quality by design principles for the accurate determination of esculin in its bulk form
Patil S, et al.
Future Journal of Pharmaceutical Sciences, 9, 69-69 (2023)
Extraction of pesticides from soil using direct-immersion SPME LC-Tips followed by GC-MS/MS: Evaluation and proof-of-concept
Brinco J, et al.
Journal of Chromatography A, 1735, 465295-465295 (2024)



Global Trade Item Number

SKUGTIN
MX0475-NP2004061839428539
MX0475-3804061839428508
MX0475-3104061839428478
MX0475P-104061839428706
MX0475-104061839428454
MX0475-3704061839428492
MX0475P-604061839428713

Questions

Reviews

No rating value

Active Filters