Skip to Content
Merck

Skip To

1494079

USP

Pancreatin lipase

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Pancreatin Lipase

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

CAS Number:
MDL number:
NACRES:
NA.24
UNSPSC Code:
41116107
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

form

solid

manufacturer/tradename

USP

application(s)

USP Biologics
pharmaceutical (small molecule)

format

neat

General description

Pancreatin Lipase is a lipolytic enzyme obtained from porcine pancreas. It catalyzes the hydrolysis of dietary triglycerides into monoglycerides and free fatty acids, aiding in lipid digestion. This enzyme is used in the treatment of exocrine pancreatic insufficiency.

The USP biologics proteins category provides critical standards and reference materials necessary for the development and manufacturing of therapeutic proteins. These proteins, which can be naturally derived or genetically engineered, are essential in treating various diseases and conditions, ensuring that they meet quality, safety, and efficacy standards.

The United States Pharmacopeia (USP) provides quality standards for biologics to ensure their safety, efficacy, and quality throughout the manufacturing process. These standards assist manufacturers in adhering to regulatory requirements and help safeguard public health by reducing risks associated with biologics.

Application

Pancreatin lipase USP Reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Pancreatin Tablets
  • Pancreatin
  • Pancrelipase Delayed-Release Capsules
  • Pancrelipase Tablets
  • Pancrelipase Capsules
  • Pancrelipase

Other Notes

One USP Unit of lipase activity is contained in the amount of pancreatin that liberates 1.0 µEq of acid per minute at a pH of 9.0 and 37° under the conditions of the assay for lipase activity.
Sales restrictions may apply.
This product is intended for use as a reference standard in pharmaceutical research and development. This product is for test and assay use only. It is not tested for animal or human consumption, clinical testing, or therapeutic use.
This product is part of the USP Biologics program.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1.07133P3292P1750
grade

pharmaceutical primary standard

grade

-

grade

-

grade

-

manufacturer/tradename

USP

manufacturer/tradename

-

manufacturer/tradename

-

manufacturer/tradename

-

application(s)

USP Biologics
pharmaceutical (small molecule)

application(s)

pharmaceutical
solid formulation

application(s)

-

application(s)

-

form

solid

form

solid

form

powder

form

powder

format

neat

format

-

format

-

format

-


pictograms

Health hazardExclamation mark

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters