Skip to Content
Merck
All Photos(3)

Key Documents

P1750

Sigma-Aldrich

Pancreatin from porcine pancreas

4 × USP specifications

Synonym(s):

Pancreatin from hog pancreas

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

Agency

USP (specifications)

form

powder

specific activity

4 × USP specifications

contains

lactose or sucrose as extender (The sucrose used might contain up to 3.25% starch.)

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Pancreatin from porcine pancreas has been used:
  • for a study to assess the treatment of steatorrhea by lipase supplementation therapy
  • for a study to investigate treatment options for pancreatic diabetes in patients experiencing the decompensated stage of chronic pancreatitis
  • to safely and effectively remove formalin-fixed tissues from arterial grafts without causing structural damage and loss in fiber integrity
  • to assess cleavage by digestive enzymes.
  • for in vitro digestibility analysis and to test the sensitivities of cellulolytic bacteria inhibitors.
  • along with amyloglucosidase for the in vitro digestion of starch in food samples.

Biochem/physiol Actions

Pancreatin contains enzymatic components including trypsin, amylase and lipase, ribonuclease, and protease, produced by the exocrine cells of the porcine pancreas. This combination of enzymes allows it to hydrolyze proteins, starch and fats. Pancreatin will convert not less than 25 times its weight of potato starch into soluble carbohydrates in 5 minutes in water at 40°C, will digest not less than 25 times its weight of casein in 60 minutes at pH 7.5 at 40°C and will release not less than microequivalents of acid per min per mg pancreatin from olive oil at pH 9.0 at 37°C.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Peter Layer et al.
Pancreas, 26(1), 1-7 (2002-12-25)
Treatment of steatorrhea by lipase supplementation therapy has become more successful in the last decade due to better understanding of the physiology and pathophysiology of the digestive process. Porcine lipase has been the therapeutic standard for several decades and will
David C Whitcomb et al.
Digestive diseases and sciences, 52(1), 1-17 (2007-01-06)
A primary function of the pancreas is to produce digestive enzymes that are delivered to the small intestine for the hydrolysis of complex nutrients. Much of our understanding of digestive enzymes comes from studies in animals. New technologies and the
Cunxi Wang et al.
Regulatory toxicology and pharmacology : RTP, 71(2), 164-173 (2014-12-30)
DroughtGard maize was developed through constitutive expression of cold shock protein B (CSPB) from Bacillus subtilis to improve performance of maize (Zea mays) under water-limited conditions. B. subtilis commonly occurs in fermented foods and CSPB has a history of safe
Asavela Ngalavu et al.
Animals : an open access journal from MDPI, 10(4) (2020-04-17)
Feed fiber composition is usually considered as one of the factors that have an impact on digestive tract microbiota composition. The investigations on the level of fermentation and in-vitro digestibility of different fibers are not well understood. The aim of
Priscila Brigide et al.
Food & function, 10(8), 4802-4810 (2019-07-19)
The present work evaluated the effect of different processes in relation to mineral content and its bioavailability, as well as the effect of phytate and oxalate contents in biofortified beans. The following treatments were evaluated: raw beans (RB), cooked and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service