Skip to Content
Merck

Skip To

1341001

USP

Indomethacin

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

1-(4-Chlorobenzoyl)-5-methoxy-2-methyl-3-indoleacetic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C19H16ClNO4
CAS Number:
Molecular Weight:
357.79
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
497341
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

API family

indomethacin

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

COc1ccc2n(c(C)c(CC(O)=O)c2c1)C(=O)c3ccc(Cl)cc3

InChI

1S/C19H16ClNO4/c1-11-15(10-18(22)23)16-9-14(25-2)7-8-17(16)21(11)19(24)12-3-5-13(20)6-4-12/h3-9H,10H2,1-2H3,(H,22,23)

InChI key

CGIGDMFJXJATDK-UHFFFAOYSA-N

Gene Information

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Indomethacin USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Indomethacin Capsules
  • Indomethacin Extended-Release Capsules
  • Indomethacin for Injection
  • Indomethacin Oral Suspension
  • Indomethacin Sodium
  • Indomethacin Suppositories

Biochem/physiol Actions

Cyclooxygenase (COX) inhibitor that is relatively selective for COX-1.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
I0200000PHR1247I7378
manufacturer/tradename

USP

manufacturer/tradename

EDQM

manufacturer/tradename

-

manufacturer/tradename

-

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

certified reference material, pharmaceutical secondary standard

grade

-

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

-

format

neat

format

neat

format

neat

format

-

API family

indomethacin

API family

indomethacin

API family

indomethacin

API family

-

Gene Information

human ... PTGS1(5742), PTGS2(5743)

Gene Information

human ... PTGS1(5742), PTGS2(5743)

Gene Information

human ... PTGS1(5742), PTGS2(5743)

Gene Information

human ... ALB(213), ALOX5(240), ELA2(1991), GPR44(11251), IL1A(3552), PLA2G1B(5319), PTGDR(5729), PTGS1(5742), PTGS2(5743)
mouse ... Alox5(11689), Ptgdr(19214), Ptger1(19216), Ptger2(19217), Ptger3(19218), Ptger4(19219), Ptgs1(19224)
rat ... Alox5(25290), Ptgs1(24693), Ptgs2(29527)


Still not finding the right product?

Explore all of our products under Indomethacin


pictograms

Skull and crossbones

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 1 Oral

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Taco J Blokhuis et al.
Clinical orthopaedics and related research, 467(2), 526-530 (2008-09-30)
Heterotopic ossification is a well-known complication after fixation of an acetabular fracture. Indomethacin and radiation therapy are used as prophylaxis to prevent heterotopic ossification. It is unclear, however, whether either is superior, although this may relate to lack of power
Ramesh Chennupati et al.
PloS one, 9(7), e102264-e102264 (2014-07-18)
Argininosuccinate synthetase (ASS) is essential for recycling L-citrulline, the by-product of NO synthase (NOS), to the NOS substrate L-arginine. Here, we assessed whether disturbed arginine resynthesis modulates endothelium-dependent vasodilatation in normal and diabetic male mice. Endothelium-selective Ass-deficient mice (Assfl/fl/Tie2Cretg/- = Ass-KOTie2) were
Brian J Krieg et al.
Journal of pharmaceutical sciences, 103(11), 3473-3490 (2014-09-13)
Development of an oral in vivo predictive dissolution medium for acid drugs with a pKa in the physiological range (e.g., Biopharmaceutics Classification System Class IIa) requires transport analysis of the complex in vivo CO2 /bicarbonate buffering system. In this report



Global Trade Item Number

SKUGTIN
1341001-200MG04061838729835

Questions

  1. What is the recommended solvent for the compound with the product number 1341001-200MG, and what are the storage conditions?

    1 answer
    1. The recommended solvents for the compound with the product number 1341001-200MG, which is a USP product, are organic solvents such as ethanol, DMSO, and dimethyl formamide. These solvents should be purged with an inert gas. The compound is soluble in water at 0.02 mg/ml and in a 45% w/v 2-Hydroxypropyl-β-cyclodextrin solution at 3.0 mg/ml.
      For the storage conditions it is to be noted that the compound is air-sensitive and not stable in solution, thus, specific solution storage information is not available.

      Helpful?

Reviews

No rating value

Active Filters