Skip to Content
Merck
All Photos(1)

Key Documents

I0200000

Indomethacin

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

1-(4-Chlorobenzoyl)-5-methoxy-2-methyl-3-indoleacetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H16ClNO4
CAS Number:
Molecular Weight:
357.79
Beilstein:
497341
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

Agency

EP Reference Standard

API family

indomethacin

manufacturer/tradename

EDQM

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

COc1ccc2n(c(C)c(CC(O)=O)c2c1)C(=O)c3ccc(Cl)cc3

InChI

1S/C19H16ClNO4/c1-11-15(10-18(22)23)16-9-14(25-2)7-8-17(16)21(11)19(24)12-3-5-13(20)6-4-12/h3-9H,10H2,1-2H3,(H,22,23)

InChI key

CGIGDMFJXJATDK-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Indomethacin EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Cyclooxygenase (COX) inhibitor that is relatively selective for COX-1.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 1 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jason P Riley et al.
Clinical science (London, England : 1979), 125(11), 533-542 (2013-06-27)
One feature of allergic asthma, the EAR (early allergic reaction), is not present in the commonly used mouse models. We therefore investigated the mediators involved in EAR in a guinea-pig in vivo model of allergic airway inflammation. Animals were sensitized
Dongfei Liu et al.
Journal of controlled release : official journal of the Controlled Release Society, 170(2), 268-278 (2013-06-13)
Nanoparticulate drug delivery systems offer remarkable opportunities for clinical treatment. However, there are several challenges when they are employed to deliver multiple cargos/payloads, particularly concerning the synchronous delivery of small molecular weight drugs and relatively larger peptides. Since porous silicon
Taco J Blokhuis et al.
Clinical orthopaedics and related research, 467(2), 526-530 (2008-09-30)
Heterotopic ossification is a well-known complication after fixation of an acetabular fracture. Indomethacin and radiation therapy are used as prophylaxis to prevent heterotopic ossification. It is unclear, however, whether either is superior, although this may relate to lack of power
Chin-Wei Chiao et al.
Clinical science (London, England : 1979), 125(3), 131-141 (2013-03-09)
Pro-inflammatory cytokines, chemokines and ROS (reactive oxygen species) are excessively produced in endotoxaemia. However, attempting to inhibit all of these inflammatory signalling pathways at the same time in order to prevent endotoxaemia is difficult. In a previous study we observed
Séverine Mazaud-Guittot et al.
The Journal of clinical endocrinology and metabolism, 98(11), E1757-E1767 (2013-09-14)
Masculinization depends on the fetal testis. Exposure of the human fetus during pregnancy to paracetamol and/or to other mild analgesics is associated with an increased risk of cryptorchidism. We aimed to determine whether mild analgesics disrupted the morphology and endocrine

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service