Skip to Content
Merck

Skip To

1138427

USP

Clobetasol Propionate Related Compound A

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

(17R)-4′-Chloro-5′-ethyl-9-fluoro-11β-hydroxy-16β-methylspiro[androsta-1,4-diene-17,2′(3′H)-furan]-3,3′-dione

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C25H30ClFO4
CAS Number:
Molecular Weight:
448.95
UNSPSC Code:
41116107
NACRES:
NA.24
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

clobetasol

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CCC1=C(Cl)C(=O)[C@@]2(O1)[C@@H](C)C[C@H]3[C@@H]4CCC5=CC(=O)C=C[C@]5(C)[C@@]4(F)[C@@H](O)C[C@]23C

InChI

1S/C25H30ClFO4/c1-5-18-20(26)21(30)25(31-18)13(2)10-17-16-7-6-14-11-15(28)8-9-22(14,3)24(16,27)19(29)12-23(17,25)4/h8-9,11,13,16-17,19,29H,5-7,10,12H2,1-4H3/t13-,16-,17-,19-,22-,23-,24-,25-/m0/s1

InChI key

LXZPFTQONPQBQD-TUZVJILZSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Clobetasol Propionate Related Compound A USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.
Also, for use with USP monographs such as:
  • Clobetasol Propionate
  • Clobetasol Propionate Cream
  • Clobetasol Propionate Ointment
  • Clobetasol Propionate Topical Solution

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
104850613029061197302
manufacturer/tradename

USP

manufacturer/tradename

USP

manufacturer/tradename

USP

manufacturer/tradename

USP

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

format

neat

format

neat

format

neat

format

neat

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

API family

clobetasol

API family

beclometasone

API family

halobetasol

API family

diflorasone

storage temp.

2-8°C

storage temp.

-

storage temp.

2-8°C

storage temp.

-


pictograms

Health hazard

signalword

Danger

Hazard Classifications

Aquatic Chronic 4 - Repr. 1B - STOT RE 2

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
1138427-50MG04061833849606

Questions

Reviews

No rating value

Active Filters