Skip to Content
Merck

Skip To

1138405

USP

Clobetasol propionate

United States Pharmacopeia (USP) Reference Standard

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C25H32ClFO5
CAS Number:
Molecular Weight:
466.97
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

API family

clobetasol

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CCC(=O)O[C@@]1([C@@H](C)C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]12C)C(=O)CCl

InChI

1S/C25H32ClFO5/c1-5-21(31)32-25(20(30)13-26)14(2)10-18-17-7-6-15-11-16(28)8-9-22(15,3)24(17,27)19(29)12-23(18,25)4/h8-9,11,14,17-19,29H,5-7,10,12-13H2,1-4H3/t14-,17-,18-,19-,22-,23-,24-,25-/m0/s1

InChI key

CBGUOGMQLZIXBE-XGQKBEPLSA-N

Gene Information

human ... NR3C1(2908)

General description

Clobetasol propionate is a fluorinated corticosteroid. It is mainly used as topical ointment.[1] It is an anti-inflammatory agent and has been considered as vasoconstrictor in nature.[2]
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Clobetasol propionate USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Clobetasol Propionate Cream
  • Clobetasol Propionate Ointment
  • Clobetasol Propionate Topical Solution

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
BP521PHR1921Y0000559
manufacturer/tradename

USP

manufacturer/tradename

BP

manufacturer/tradename

-

manufacturer/tradename

EDQM

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

certified reference material, pharmaceutical secondary standard

grade

pharmaceutical primary standard

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical
pharmaceutical small molecule

application(s)

pharmaceutical

application(s)

pharmaceutical (small molecule)

format

neat

format

neat

format

-

format

neat

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

Gene Information

human ... NR3C1(2908)

Gene Information

human ... NR3C1(2908)

Gene Information

human ... NR3C1(2908)

Gene Information

human ... NR3C1(2908)


Still not finding the right product?

Explore all of our products under Clobetasol propionate


pictograms

Health hazard

signalword

Danger

Hazard Classifications

Aquatic Chronic 4 - Repr. 1B - STOT RE 2

target_organs

Adrenal gland,Immune system

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



J A Carruthers et al.
British medical journal, 4(5990), 203-204 (1975-10-25)
Topical application of clobetasol propionate in a strength of 0-05% in cream or ointment (Dermovate) suppressed the hypothalamic-pituitary-adrenal axis in both normal people and patients with diseased skin. In normal people the 9 am serum cortisol level was suppressed when
Marika Kamberi et al.
Journal of chromatographic science, 46(1), 23-29 (2008-01-26)
A novel sensitive high-throughput high-performance liquid chromatography assay is developed and validated for the simultaneous determination of everolimus and clobetasol propionate in pharmaceutical formulations. The chromatographic separation is achieved on a Zorbax Eclipse XDB-C18 reversed-phase column using a gradient elution
Lindsey Warino et al.
Journal of drugs in dermatology : JDD, 5(6), 527-532 (2006-06-16)
Clobetasol propionate is the most common topical therapy used for psoriasis in the US. Conventional dermatologic wisdom is that ointment preparations provide the highest potency (due to their occlusive nature and moisturizing ability) and are best suited for psoriasis. However



Global Trade Item Number

SKUGTIN
1138405-200MG04061838701503

Questions

Reviews

No rating value

Active Filters