Skip to Content
Merck

Skip To

1071508

USP

Biotin

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

D-Biotin, Bios II, Coenzyme R, Vitamin B7, Vitamin H

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C10H16N2O3S
CAS Number:
Molecular Weight:
244.31
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
86838
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

API family

biotin

manufacturer/tradename

USP

mp

231-233 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

[H][C@]12CS[C@@H](CCCCC(O)=O)[C@@]1([H])NC(=O)N2

InChI

1S/C10H16N2O3S/c13-8(14)4-2-1-3-7-9-6(5-16-7)11-10(15)12-9/h6-7,9H,1-5H2,(H,13,14)(H2,11,12,15)/t6-,7-,9-/m0/s1

InChI key

YBJHBAHKTGYVGT-ZKWXMUAHSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Biotin USP reference standard suitable for use in specified USP compendial tests and assays.

Also used to prepare standard and standard stock solutions for assay by using liquid chromatography coupled with a UV detector according to the given below monographs of United States Pharmacopeia (USP):
  • Biotin Compounded Oral Suspension
  • Biotin Tablets
  • Biotin Capsules
  • Biotin
  • Water-Soluble Vitamins Tablets

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
B030191827RES1052B-B7
grade

pharmaceutical primary standard

grade

-

grade

certified reference material, TraceCERT®

grade

-

manufacturer/tradename

USP

manufacturer/tradename

-

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

manufacturer/tradename

-

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

clinical testing
pharmaceutical (small molecule)

application(s)

-

format

neat

format

-

format

neat

format

-

mp

231-233 °C (lit.)

mp

231-233 °C (lit.)

mp

231-233 °C (lit.)

mp

231-233 °C (lit.)

API family

biotin

API family

-

API family

-

API family

-


Still not finding the right product?

Explore all of our products under Biotin


Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Biotin Tablets
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 38(5), 571-571 (2013)
Water-Soluble Vitamins Tablets
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 44(6), 5512-5512 (2020)
Biotin Capsules
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 38(5), 570-570 (2013)



Global Trade Item Number

SKUGTIN
1071508-200MG04061838674586

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters