Skip to Content
Merck
All Photos(4)

Documents

B0301

Sigma-Aldrich

Biotin

meets USP testing specifications

Synonym(s):

D-Biotin, Bios II, Coenzyme R, Vitamin B7, Vitamin H

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16N2O3S
CAS Number:
Molecular Weight:
244.31
Beilstein:
86838
EC Number:
MDL number:
UNSPSC Code:
12352201
eCl@ss:
34058011
PubChem Substance ID:
NACRES:
NA.21

biological source

synthetic (organic)

Quality Level

Agency

meets USP testing specifications

Assay

97.5-102.0%

form

powder

color

white

mp

231-233 °C (lit.)

application(s)

pharmaceutical (small molecule)

storage temp.

2-8°C

SMILES string

[H][C@]12CS[C@@H](CCCCC(O)=O)[C@@]1([H])NC(=O)N2

InChI

1S/C10H16N2O3S/c13-8(14)4-2-1-3-7-9-6(5-16-7)11-10(15)12-9/h6-7,9H,1-5H2,(H,13,14)(H2,11,12,15)/t6-,7-,9-/m0/s1

InChI key

YBJHBAHKTGYVGT-ZKWXMUAHSA-N

Looking for similar products? Visit Product Comparison Guide

Application

<ul>
<li><strong>Protein Biotinylation Studies:</strong> Advanced strategies for biotinylation site characterization in proteins are being developed using biotin, improving the precision of protein modification techniques critical for biochemical research and drug development (Zhu, 2024).</li>
</ul>

Biochem/physiol Actions

Essential vitamin that is important for amino acid and energy metabolism, and fatty acid synthesis. It is a prosthetic group in four mammalian carboxylase families and facilitates the binding and transfer of carbon dioxide.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tazu Aoki et al.
Neuron, 78(5), 881-894 (2013-05-21)
The encoding of long-term associative memories for learned behaviors is a fundamental brain function. Yet, how behavior is stably consolidated and retrieved in the vertebrate cortex is poorly understood. We trained zebrafish in aversive reinforcement learning and measured calcium signals
Naruo Nikoh et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(28), 10257-10262 (2014-07-02)
Obligate insect-bacterium nutritional mutualism is among the most sophisticated forms of symbiosis, wherein the host and the symbiont are integrated into a coherent biological entity and unable to survive without the partnership. Originally, however, such obligate symbiotic bacteria must have
Tuan V Bui et al.
Neuron, 78(1), 191-204 (2013-04-16)
Accurate motor performance depends on the integration in spinal microcircuits of sensory feedback information. Hand grasp is a skilled motor behavior known to require cutaneous sensory feedback, but spinal microcircuits that process and relay this feedback to the motor system
Konstantinos Ampatzis et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 33(26), 10875-10886 (2013-06-28)
In vertebrates, spinal circuits drive rhythmic firing in motoneurons in the appropriate sequence to produce locomotor movements. These circuits become active early during development and mature gradually to acquire the flexibility necessary to accommodate the increased behavioral repertoire of adult
Stuart Trenholm et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 33(37), 14927-14938 (2013-09-13)
Recently, we demonstrated that gap junction coupling in the population of superior coding ON-OFF directionally selective ganglion cells (DSGCs) genetically labeled in the Hb9::eGFP mouse retina allows the passage of lateral anticipatory signals that help track moving stimuli. Here, we

Articles

Importance and uses of biotin in serum-free eukaryotic, including hybridoma and Chinese Hamster Ovary (CHO) cell, cultures

Importance and uses of biotin in serum-free eukaryotic, including hybridoma and Chinese Hamster Ovary (CHO) cell, cultures

Importance and uses of biotin in serum-free eukaryotic, including hybridoma and Chinese Hamster Ovary (CHO) cell, cultures

Importance and uses of biotin in serum-free eukaryotic, including hybridoma and Chinese Hamster Ovary (CHO) cell, cultures

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service