Skip to Content
Merck

Skip To

1021204

USP

4-Aminophenol

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

4-Hydroxyaniline

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Linear Formula:
H2NC6H4OH
CAS Number:
Molecular Weight:
109.13
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
385836
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

API family

paracetamol, acetaminophen, mesalazine

manufacturer/tradename

USP

mp

185-189 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

Nc1ccc(O)cc1

InChI

1S/C6H7NO/c7-5-1-3-6(8)4-2-5/h1-4,8H,7H2

InChI key

PLIKAWJENQZMHA-UHFFFAOYSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets, has been developed and issued under the Authority of the issuing Pharmacopoeia.

For further information and support please go to the website of the issuing Pharmacopoeia.4-Aminophenol is the principal degradation product of acetaminophen, formed as an impurity during its hydrolysis.

Application

This USP reference standard is intended for use in specified quality tests and assays.
Also used to prepare standard and system suitability solutions to test for the presence of 4-aminophenol as an impurity in various pharmaceutical dosage forms of acetaminophen, according to the United States Pharmacopeia (USP) monographs:
  • Mesalamine
  • Acetaminophen Tablets
  • Acetaminophen Oral Suspension
  • Tramadol Hydrochloride and Acetaminophen Tablets

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
PHR1148A713288.00421
manufacturer/tradename

USP

manufacturer/tradename

-

manufacturer/tradename

-

manufacturer/tradename

-

grade

pharmaceutical primary standard

grade

certified reference material, pharmaceutical secondary standard

grade

-

grade

-

format

neat

format

neat

format

-

format

-

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

-

application(s)

-

mp

185-189 °C (lit.)

mp

185-189 °C (lit.)

mp

185-189 °C (lit.)

mp

190 °C

API family

paracetamol, acetaminophen, mesalazine

API family

mesalazine, paracetamol, acetaminophen

API family

-

API family

-


Still not finding the right product?

Explore all of our products under 4-Aminophenol


signalword

Warning

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Muta. 2 - Skin Sens. 1 - STOT RE 2

target_organs

Kidney

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Acetaminophen Oral Suspension
USP43-NF38: United States Pharmacopeia and National Formulary
United States Pharmacopeia, 42(6), 42-42 (2021)
Mesalamine
USP43-NF38: United States Pharmacopeia and National Formulary
United States Pharmacopeia, 40(6), 2800-2800 (2020)
Tramadol Hydrochloride and Acetaminophen Tablets
USP43-NF38: United States Pharmacopeia and National Formulary
United States Pharmacopeia, 42(1), 4450-4450 (2017)



Global Trade Item Number

SKUGTIN
1021204-100MG04061838669261

Questions

Reviews

No rating value

Active Filters