Skip to Content
Merck
All Photos(1)

Key Documents

1003009

USP

Acetaminophen

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Acetaminophen, 4′-Hydroxyacetanilide, 4-Acetamidophenol, N-(4-Hydroxyphenyl)acetamide, N-Acetyl-4-aminophenol, APAP, Paracetamol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CONHC6H4OH
CAS Number:
Molecular Weight:
151.16
Beilstein:
2208089
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

acetaminophen, paracetamol

manufacturer/tradename

USP

mp

168-172 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

CC(=O)Nc1ccc(O)cc1

InChI

1S/C8H9NO2/c1-6(10)9-7-2-4-8(11)5-3-7/h2-5,11H,1H3,(H,9,10)

InChI key

RZVAJINKPMORJF-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Acetaminophen USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.
Also, for use with USP monographs such as:
  • Acetaminophen Tablets
  • Acetaminophen Capsules
  • Acetaminophen Suppositories
  • Acetaminophen Oral Suspension
  • Acetaminophen Oral Solution
  • Acetaminophen and Caffeine Tablets
  • Acetaminophen Extended-Release Tablets

Analgesic.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

364.3 °F - Pensky-Martens closed cup

Flash Point(C)

184.6 °C - Pensky-Martens closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Acetaminophen
USP43-NF38: United States Pharmacopeia and National Formulary
United States Pharmacopeia, 39(6), 38-38 (2020)
Acetaminophen capsules
USP43-NF38: United States Pharmacopeia and National Formulary
United States Pharmacopeia, 45(3), 40-40 (2014)
Acetaminophen tablets
USP43-NF38: United States Pharmacopeia and National Formulary
United States Pharmacopeia, 43(3), 45-45 (2021)
Acetaminophen Suppositories
USP43-NF38: United States Pharmacopeia and National Formulary
United States Pharmacopeia, 41(5), 44-44 (2016)
Karel Allegaert et al.
Archives of disease in childhood, 98(6), 462-466 (2013-04-23)
There remains a need for alternative medical treatments for patent ductus arteriosus (PDA) closure in extreme preterm neonates because of therapeutic failure and adverse effects associated with non-selective cyclo-oxygenase inhibitors. Reports of an association between paracetamol exposure and PDA closure

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service