Skip to Content
Merck
All Photos(1)

Key Documents

1012065

USP

Acyclovir

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Acycloguanosine, 9-[(2-Hydroxyethoxy)methyl]guanine, Acyclovir

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H11N5O3
CAS Number:
Molecular Weight:
225.20
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

acyclovir

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

NC1=Nc2c(ncn2COCCO)C(=O)N1

InChI

1S/C8H11N5O3/c9-8-11-6-5(7(15)12-8)10-3-13(6)4-16-2-1-14/h3,14H,1-2,4H2,(H3,9,11,12,15)

InChI key

MKUXAQIIEYXACX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Acyclovir is a nucleoside analogue antiviral drug. It is a highly potent inhibitor of herpes simplex virus.[1] It is related to cytarabine, idoxuridine, trifluridine and vidarabine.[2]

Application

Acyclovir USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Acyclovir Capsules
  • Acyclovir for Injection
  • Acyclovir Injection
  • Acyclovir Ointment
  • Acyclovir Oral Suspension
  • Acyclovir Tablets

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D M Richards et al.
Drugs, 26(5), 378-438 (1983-11-01)
Acyclovir (aciclovir) is a nucleoside analogue antiviral drug related to cytarabine, idoxuridine, trifluridine and vidarabine. In common with these earlier antivirals, acyclovir is active against some members of the herpesvirus group of DNA viruses. The efficacy of topical acyclovir has
Ahlem Ben Sassi et al.
The Indian journal of medical research, 127(2), 183-192 (2008-04-12)
Tunisian Chrysanthemum species are known to have medicinal activity and some of the species are used in traditional medicine. We have earlier shown the use of C. trifurcatum flowerheads in Tunisian traditional medicine to treat constipation. In the present study
G B Elion
The American journal of medicine, 73(1A), 7-13 (1982-07-20)
Acyclovir, an acrylic purine nucleoside analog, is a highly potent inhibitor of herpes simplex virus (HSV), types 1 and 2, and varicella zoster virus, and has extremely low toxicity for the normal host cells. This selectivity is due to the
Derivative spectrophotometric method for determination of acyclovir in polymeric nanoparticles.
Tavares, Guilherme Diniz, et al.
Quimica nova, 35.1, 203-206 (2012)
Suneel Chilukuri et al.
Dermatologic clinics, 21(2), 311-320 (2003-05-22)
In immunocompetent patients, HSV is controlled rapidly by the human host's immune system, and recurrent lesions are small and short lived. When treated with antiviral agents, these patients rarely develop resistance to these drugs. In contrast immunocompromised patients might not

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service