Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
grade
pharmaceutical primary standard
agency
USP
description
API family: acyclovir
API family
acyclovir
manufacturer/tradename
USP
application(s)
pharmaceutical (small molecule)
format
neat
SMILES string
CC(=O)OCCOCn1cnc2C(=O)NC(N)=Nc12
InChI
1S/C10H13N5O4/c1-6(16)19-3-2-18-5-15-4-12-7-8(15)13-10(11)14-9(7)17/h4H,2-3,5H2,1H3,(H3,11,13,14,17)
InChI key
DMHAXLGAKQREIL-UHFFFAOYSA-N
General description
Application
- Acyclovir
- Valacyclovir Hydrochloride
Analysis Note
Other Notes
1 of 1
This Item | |||
|---|---|---|---|
| manufacturer/tradename USP | manufacturer/tradename - | manufacturer/tradename - | manufacturer/tradename - |
| grade pharmaceutical primary standard | grade certified reference material, pharmaceutical secondary standard | grade certified reference material, pharmaceutical secondary standard | grade certified reference material, pharmaceutical secondary standard |
| application(s) pharmaceutical (small molecule) | application(s) pharmaceutical small molecule | application(s) pharmaceutical | application(s) pharmaceutical |
| format neat | format - | format - | format - |
| API family acyclovir | API family - | API family - | API family - |
| description API family: acyclovir | description - | description - | description - |
Still not finding the right product?
Explore all of our products under Acyclovir Related Compound A
signalword
Warning
hcodes
Hazard Classifications
Muta. 2
Storage Class
11 - Combustible Solids
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



