Skip to Content
Merck

Skip To

U5378

Urea

powder, BioReagent, Molecular Biology, suitable for cell culture

Urea

Synonym(s):

Carbamide, Carbonyldiamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Linear Formula:
NH2CONH2
CAS Number:
Molecular Weight:
60.06
UNSPSC Code:
12352111
NACRES:
NA.31
PubChem Substance ID:
EC Number:
200-315-5
Beilstein/REAXYS Number:
635724
MDL number:
Assay:
≥98%
Grade:
Molecular Biology
Technique(s):
cell culture | mammalian: suitable
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

Molecular Biology

Quality Level

product line

BioReagent

assay

≥98%

form

powder

technique(s)

cell culture | mammalian: suitable

mp

132-135 °C (lit.)

solubility

H2O: soluble 480 mg/mL, clear, colorless (8M)

density

1.335 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤5 ppm

cation traces

Cu: ≤1 ppm, Fe: ≤5 ppm, Pb: ≤2 ppm

absorption

≤0.06 at 260 at 6 M, ≤0.06 at 280 at 6 M

foreign activity

DNase, NICKase, RNase and protease, none detected

SMILES string

NC(N)=O

InChI

1S/CH4N2O/c2-1(3)4/h(H4,2,3,4)

InChI key

XSQUKJJJFZCRTK-UHFFFAOYSA-N

Gene Information

human ... CA1(759), CA2(760)
rat ... Ppm1a(24666)

Application

Used for the denaturation of proteins and as a mild solubilization agent for insoluble or denatured proteins. Useful for renaturing proteins from samples already denatured with 6 M guanidine chloride such as inclusion bodies. May be used with guanidine hydrochloride and dithiothreitrol (DTT) in the refolding of denatured proteins into their native or active form.
  • Urea has been used in protein resuspension buffer and to denature proteins.
  • It has been used for the isolation of proteins via urea-containing polyacrylamide gels.
  • It has been used to study the conformation of unfolded proteins under different concentrations of urea and different pH.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
U0631U125015604
technique(s)

cell culture | mammalian: suitable

technique(s)

-

technique(s)

-

technique(s)

-

grade

Molecular Biology

grade

-

grade

-

grade

-

assay

≥98%

assay

98.5-101.5%

assay

≥99.5%

assay

99.0-100.5%, 99.0-101.0% (calc. on dry substance)

Quality Level

200

Quality Level

200

Quality Level

300

Quality Level

200

solubility

H2O: soluble 480 mg/mL, clear, colorless (8M)

solubility

H2O: 5 M at 20 °C, clear, colorless

solubility

H2O: 8 M

solubility

H2O: soluble 480 g/L at 20 °C

form

powder

form

powder, crystals or pellets

form

pellets

form

solid


Still not finding the right product?

Explore all of our products under Urea


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content

Product Information Sheet


Labeling and identification of direct kinase substrates.
Carlson SM and White FM
Science Signaling, 5, pl3-pl3 (2012)
Construction and transfection of a ribozyme targeting human caspase-3.
Cronin D et al.
Anticancer Research, 24, 425-425 (2004)
Structure and properties of native and unfolded lysing enzyme from T. harzianum: Chemical and pH denaturation.
Bey H et al.
International Journal of Biological Macromolecules, 92, 860-860 (2016)



Global Trade Item Number

SKUGTIN
U5378-500G04061835550449
U5378-5KG04061835550456
U5378-100G04061835522675
U5378-1KG04061835550432

Questions

1–3 of 3 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  3. what is storage temperature for U5378?

    1 answer
    1. This product may be stored at room temperature. For products with no specified storage temperature given on the label or product page, ambient storage may be assumed.

      Helpful?

Reviews

Active Filters

  1. 1 Ratings-only review