Skip to Content
Merck
All Photos(1)

Key Documents

15604

Sigma-Aldrich

Urea

puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)

Synonym(s):

Carbamide, Carbonyldiamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2CONH2
CAS Number:
Molecular Weight:
60.06
Beilstein:
635724
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21

grade

puriss.

Quality Level

Assay

99.0-100.5%
99.0-101.0% (calc. on dry substance)

form

solid

quality

meets analytical specification of Ph. Eur., BP, USP

impurities

alkalic reac. substances, complies
≤0.001% heavy metals (as Pb)
≤0.04% insoluble in ethanol
≤0.05% ammonium (NH4)
≤0.1% biuret(carbamylurea)

ign. residue

≤0.1% (as SO4)

loss

≤1.0% loss on drying, 105 °C, 1 h

mp

132-135 °C (lit.)
132-135 °C

solubility

H2O: soluble 480 g/L at 20 °C

density

1.335 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤10 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Fe: ≤0.5 mg/kg

suitability

in accordance for appearance of solution
in accordance for identity

functional group

amine

SMILES string

NC(N)=O

InChI

1S/CH4N2O/c2-1(3)4/h(H4,2,3,4)

InChI key

XSQUKJJJFZCRTK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Urea is a colorless, odorless solid. It has prism-like crystals. It forms insoluble nitrate salts with nitric acid and oxalate salts with oxalic acid. On reaction with alkanes or substituted alkanes, it affords crystal-lattice inclusion compounds.
Urea can be prepared from ammonia and carbon-dioxide, via Haber-Bosch process. It can also be used to prepare urea-formaldehyde resins, via condensation reaction.

Application

Urea has been used in the following studies:
  • Synthesis of SnO2 nanoparticles.
  • Synthesis of urethanes, ureides and semicarbazides.
  • Denaturation of protein isolated from Saccharomyces cerevisiae Y15696 strain
  • Protein electrophoretic studies.
Used for the denaturation of proteins and as a mild solubilization agent for insoluble or denatured proteins. Useful for renaturing proteins from samples already denatured with 6 M guanidine chloride such as inclusion bodies. May be used with guanidine hydrochloride and dithiothreitrol (DTT) in the refolding of denatured proteins into their native or active form.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 7

1 of 7

Nova I and Tronconi E et al.
Urea-SCR Technology for deNOx after treatment of Diesel Exhausts (2014)
Braun D, et al. et al.
Polymer Synthesis: Theory and Practice: Fundamentals, Methods, Experiments ; with 31 Tables (2001)
Joost P J J Hegmans et al.
The American journal of pathology, 164(5), 1807-1815 (2004-04-28)
Exosomes are small membrane vesicles secreted into the extracellular compartment by exocytosis. Tumor exosomes may be involved in the sampling of antigens to antigen presenting cells or as decoys allowing the tumor to escape immune-directed destruction. The proteins present in
Genna L Andrews et al.
Analytical chemistry, 83(13), 5442-5446 (2011-05-31)
The TripleTOF 5600 System, a hybrid quadrupole time-of-flight mass spectrometer, was evaluated to explore the key figures of merit in generating peptide and protein identifications that included spectral acquisition rates, data quality, proteome coverage, and biological depth. Employing a Saccharomyces
Eagleson M.
Concise Encyclopedia Chemistry, 1137-1137 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service