Skip to Content
Merck

Skip To

SML2011

Ciclopirox

≥98% (HPLC)

Synonym(s):

Ciclopirox, 6-Cyclohexyl-1-hydroxy-4-methyl-2(1H)-pyridinone, HOE 296b, 6-Cyclohexyl-1-hydroxy-4-methyl-2(1H)-pyridone

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C12H17NO2
CAS Number:
Molecular Weight:
207.27
UNSPSC Code:
12352200
NACRES:
NA.77
MDL number:
Assay:
≥98% (HPLC)
Form:
powder
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

shipped in

ambient

storage temp.

2-8°C

SMILES string

ON1C(C2CCCCC2)=CC(C)=CC1=O

InChI

1S/C12H17NO2/c1-9-7-11(13(15)12(14)8-9)10-5-3-2-4-6-10/h7-8,10,15H,2-6H2,1H3

InChI key

SCKYRAXSEDYPSA-UHFFFAOYSA-N

Biochem/physiol Actions

Ciclopirox is a chemical compound belonging to the class of hydroxypyridones. It is found active against dermatophytes, yeasts, molds, some bacterias and also azole-resistant Candida species. Unlike other antifungal agents that act for ergosterol inhibition, ciclopirox has a different action. It targets metal-dependent enzymes, which degrades fungal cell peroxides. This unique action of ciclopirox provides less chance for resistance in pathogenic fungi. This compound is used in topical formulations specifically for nails and skin.[1]
Ciclopirox is an antifungal and cell-permeable iron-chelating agent. It has been found to inhibit multiple enzymes and signaling pathways including prolyl hydroxylase 2 (PHD2), eukaryotic translation initiation factor 5A (eIF5A), Wnt/β-catenin, hypoxia-inducible factor-1α (HIF-1 α)/vascular endothelial growth factor (VEGF), vascular endothelial growth factor receptor 3 (VEGFR-3), mammalian target of rapamycin (mTOR), and cyclin dependent kinases (CDKs). Ciclopirox has been shown to have anti-viral and anticancer activity in addition to its antifungal activity. Ciclopirox has also been shown to counteract glucotoxicity in diabetic mice in a p21-dependent manner.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
PHR19201134018SML1276
form

powder

form

-

form

-

form

powder

assay

≥98% (HPLC)

assay

-

assay

-

assay

≥98% (HPLC)

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

solubility

DMSO: 2 mg/mL, clear

solubility

-

solubility

-

solubility

DMSO: 10 mg/mL, clear

color

white to beige

color

-

color

-

color

white to beige

shipped in

ambient

shipped in

-

shipped in

-

shipped in

-


Still not finding the right product?

Explore all of our products under Ciclopirox


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class

11 - Combustible Solids

wgk

WGK 3



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library







Global Trade Item Number

SKUGTIN
SML2011-5MG04061832630748
SML2011-25MG04061832630731
SML2011-50MG04061835302840
SML2011-100MG04061835303045

Questions

Reviews

No rating value

Active Filters