
Ciclopirox

Synonym(s):
Ciclopirox, 6-Cyclohexyl-1-hydroxy-4-methyl-2(1H)-pyridone
Empirical Formula (Hill Notation):
C12H17NO2
CAS Number:
Molecular Weight:
207.27
Products
Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Purity
Agency
Format
Research Area
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-4 of 4