Skip to Content
Merck

PP0200

GlycoProfile I, In-Gel Deglycosylation Kit

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352200
EC Number:
232-615-7
NACRES:
NA.32
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


shelf life

≥1 yr at 2‑8 °C

Quality Segment

storage temp.

2-8°C

Application

The GlycoProfile Enzymatic In-gel N-Deglycosylation Kit is optimized to provide a convenient, reproducible method to N-deglycosylate and digest protein samples from one dimensional (1D) and two dimensional (2D) polyacrylamide gel pieces for subsequent mass spectrometry or HPLC analysis.
This procedure is suitable for Coomassie® Brilliant Blue and colloidal Coomassie stained gels. Silver stained gels may also be used if properly destained.

Biochem/physiol Actions

The kit includes Proteomics Grade PNGase F and trypsin enzymes necessary for N-linked deglycosylation and tryptic digestion, respectively. The samples can then be desalted and concentrated for analysis by MALDI-TOF MS or electrospray MS with subsequent database searching.

Preparation Note

Each kit contains reagents sufficient to deglycosylate and digest up to 10 samples.

Legal Information

GlycoProfile is a trademark of Sigma-Aldrich Co. LLC


Kit Components Only

Product No.
Description

  • Acetonitrile, ≥99.93%, BioSyn® 50 mL

Kit Components Also Available Separately

Product No.
Description
SDS & Pricing

  • PNGase F from Elizabethkingia meningoseptica, BioReagent, ≥95% (SDS-PAGE) 50 U
    SDS

  • Trypsin from porcine pancreas, Proteomics Grade, BioReagent, Dimethylated 20 μg
    SDS

  • Trypsin Solubilization Reagent
    SDS

  • Invertase Glycoprotein Standard, BioReagent, from Saccharomyces cerevisiae
    SDS

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

3 - Flammable liquids

flash_point_f

35.6 °F

flash_point_c

2.0 °C

wgk

WGK 3



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library






Global Trade Item Number

SKUGTIN
PP0200-1KT04061836686888