Skip to Content
Merck
All Photos(1)

Documents

T6567

Sigma-Aldrich

Trypsin from porcine pancreas

Proteomics Grade, BioReagent, Dimethylated

Synonym(s):

Porcine Trypsin, Trypsin for Mass Spectropetry

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.56

biological source

Porcine pancreas

product line

BioReagent

solubility

1 mM HCl: soluble 1 mg/mL, clear, colorless

shipped in

wet ice

storage temp.

2-8°C

General description

Trypsin is a serine protease that is usually used in biochemistry and biology as an important enzymatic reagent. This method produces a highly purified trypsin product suitable for proteomics research. Proteomics Grade ideal for use in both solution and in-gel tryptic digestions. Trypsin, a serine protease, is present in the digestive system of several vertebrates.

Application

Trypsin from porcine pancreas is used for the following applications:
  • In-gel protein digestion and MALDI-TOF mass spectrometry analysis
  • Electrospray Ionization Mass Spectrometry (ESI-MS) analysis
  • Surface proteome profiling of L. plantarum
  • Gel Filtration, Ultracentrifugation, and Rotary Shadowing Electron Microscopy
  • Mass spectrometry

Biochem/physiol Actions

Trypsin is routinely used in proteomics research for peptide mapping and protein sequence work, due to its highly specific cleavage resulting in a limited number of tryptic peptides. It hydrolyzes peptide bonds specifically at the carboxyl side of arginine and lysine residues.. The enzyme also exhibits esterase and amidase activities. Trypsin acts as a cell culture tool. It is used to hydrolyze allergenic proteins to produce hypoallergenic milk in industries.

related product

Product No.
Description
Pricing

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cristina C Santini et al.
Scientific reports, 9(1), 10865-10865 (2019-07-28)
Small molecule inhibitors of BRAF and MEK have proven effective at inhibiting tumor growth in melanoma patients, however this efficacy is limited due to the almost universal development of drug resistance. To provide advanced insight into the signaling responses that
Shaoming Mao et al.
Molecular bioSystems, 7(5), 1660-1677 (2011-03-09)
The solventogenic bacterium Clostridium acetobutylicum is the most important species of Clostridium used in the fermentation industry. However, the intolerance to butanol hampers the efficient production of solvents. Butanol toxicity has been attributed to the chaotropic effect on the cell
Families of serine peptidases.
N D Rawlings et al.
Methods in enzymology, 244, 19-61 (1994-01-01)
Daniela M Remus et al.
Journal of bacteriology, 195(3), 502-509 (2012-11-24)
Sortases are transpeptidases that couple surface proteins to the peptidoglycan of Gram-positive bacteria, and several sortase-dependent proteins (SDPs) have been demonstrated to be crucial for the interactions of pathogenic and nonpathogenic bacteria with their hosts. Here, we studied the role
Masayuki Shimoda et al.
Cell transplantation, 21(2-3), 465-471 (2012-07-17)
Porcine islets are considered to be a promising resource for xenotransplantation. However, it is difficult to isolate porcine islets because of the marked fragility and rapid dissociation. Endogenous trypsin is one of the main factors to damage islets during the

Articles

Evaluation of Recombinant, Chemically Treated Trypsin in Proteomics and Protein Characterization Assays

Get better detection and quantification of proteases with this high-sensitivity red protease detection assay.

Get better detection and quantification of proteases with this high-sensitivity red protease detection assay.

Get better detection and quantification of proteases with this high-sensitivity red protease detection assay.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service