Skip to Content
Merck

Skip To

P9130

Pyridoxal hydrochloride

≥99% (HPLC)

Synonym(s):

3-Hydroxy-5-(hydroxymethyl)-2-methyl-4-pyridinecarboxaldehyde hydrochloride, PL HCl

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C8H9NO3 · HCl
CAS Number:
Molecular Weight:
203.62
UNSPSC Code:
12352205
NACRES:
NA.79
PubChem Substance ID:
EC Number:
200-602-5
Beilstein/REAXYS Number:
3656994
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

synthetic (organic)

Quality Segment

assay

≥99% (HPLC)

form

powder

technique(s)

HPLC: suitable

color

white to off-white

mp

173 °C (dec.) (lit.)

storage temp.

−20°C

SMILES string

Cl[H].[H]C(=O)c1c(CO)cnc(C)c1O

InChI

1S/C8H9NO3.ClH/c1-5-8(12)7(4-11)6(3-10)2-9-5;/h2,4,10,12H,3H2,1H3;1H

InChI key

FCHXJFJNDJXENQ-UHFFFAOYSA-N

Application

For the labeling of amino acids and their detection in picomolar amounts; Coenzymes and Cofactors, vol. 1: Vitamin B6, Pyridoxal Phosphate
Pyridoxal hydrochloride has been used as a calibrant for liquid chromatography of food samples. It has also been used as a component of arginine decarboxylase broth and lysine decarboxylase broth (LDB) for culturing lactic acid bacteria (LAB) and food-borne pathogens (FBP), respectively.

Biochem/physiol Actions

Pyridoxal hydrochloride interacts with cysteine of cathepsins and inhibits their activity. It elicits protective function in cornu Ammonis (CA) 1 neurons during ischemia.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
93759P6155271748
technique(s)

HPLC: suitable

technique(s)

-

technique(s)

cell culture | insect: suitable, cell culture | mammalian: suitable

technique(s)

-

assay

≥99% (HPLC)

assay

≥99.5% (T)

assay

≥99% (HPLC)

assay

99%

biological source

synthetic (organic)

biological source

synthetic

biological source

-

biological source

synthetic (Organic)

form

powder

form

powder

form

powder

form

powder

storage temp.

−20°C

storage temp.

-

storage temp.

−20°C

storage temp.

2-8°C

Quality Level

200

Quality Level

300

Quality Level

200

Quality Level

200


Still not finding the right product?

Explore all of our products under Pyridoxal hydrochloride


Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library







Global Trade Item Number

SKUGTIN
P9130-25G04061834402374
P9130-5G04061835548026
P9130-500MG04061834402381
P9130-100G04061834402350
P9130-1G04061834402367

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters