Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Pyridoxal hydrochloride
Synonym(s):
3-Hydroxy-5-(hydroxymethyl)-2-methyl-4-pyridinecarboxaldehyde hydrochloride, PL HCl
Empirical Formula (Hill Notation):
C
8
H
9
NO
3
· HCl
CAS Number:
65-22-5
Molecular Weight:
203.62
EC Number:
200-602-5
Products
Product Category
vitamins (4)
Brand
Sigma-Aldrich (4)
Biological Source
Color
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Application
Technique
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
P9130
Pyridoxal hydrochloride,
≥99% (HPLC)
Expand
View Pricing
P6155
Pyridoxal hydrochloride,
BioReagent, suitable for cell culture, suitable for insect cell culture
Expand
View Pricing
271748
Pyridoxal hydrochloride,
99%
Expand
View Pricing
93759
Pyridoxal hydrochloride,
≥99.5% (T)
Expand
View Pricing