No, this product does not contain a His-tag. See the link below for additional product information: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/394/024/p7582dat.pdf
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Pack Size | SKU | Availability | Price |
|---|
About This Item
Form:
liquid
Mol wt:
~49 kDa
Pricing and availability is not currently available.
General description
Amino-terminal FLAG-BAP™ Fusion Protein is a 467 amino acid N-terminal FLAG fusion protein of E. coli bacterial alkaline phosphatase (BAP) with a calculated molecular mass of 49.3kDa. The N-terminal FLAG-BAP Fusion Protein migrates as a 45−55kDa band by SDS-PAGE depending on electrophoresis conditions. FLAG-BAP™ Fusion Protein has biotechnological applications and is used in the screening of recombinant fusion proteins.
The Amino-terminal FLAG-BAP Fusion Protein is a control prorein used to confirm the funtional integrity of the anti-FLAG M1 and M2 monoclonal antibodies in applications such as western blotting, immunoprecipitation, ELISA, electron microscopy, and FACS.
Application
Amino-terminal FLAG-BAP™ Fusion Protein has been used as a standard in enzyme-linked immunosorbent assay (ELISA) for osteopontin quantification and in western blotting based quantification of Flag-Wnt8[1] and Drosophila melanogaster transcription factor interactome.
Learn more product details in our FLAG® application portal.
Physical form
Protein is supplied in 10 mM Tris, 120 mM NaCl,
0.05 mM ZnCl2 in 50% glycerol, pH 8.0.
0.05 mM ZnCl2 in 50% glycerol, pH 8.0.
Other Notes
Control protein
Legal Information
FLAG is a registered trademark of Merck KGaA, Darmstadt, Germany
FLAG-BAP is a trademark of Sigma-Aldrich Co. LLC
1 of 1
This Item | |||
|---|---|---|---|
| mol wt ~49 kDa | mol wt ~49 kDa | mol wt 1012.97 Da | mol wt ~49 kDa |
| form liquid | form liquid | form lyophilized powder | form - |
| shipped in dry ice | shipped in dry ice | shipped in wet ice | shipped in dry ice |
| storage temp. −20°C | storage temp. −20°C | storage temp. 2-8°C | storage temp. −20°C |
| Quality Level 200 | Quality Level 200 | Quality Level 200 | Quality Level 200 |
Storage Class
10 - Combustible liquids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
Instructions
BAP-fusion: A versatile molecular probe for biotechnology research
Biotechnology: Research, Technology and Applications, 327-345 (2008)
A Comprehensive Drosophila melanogaster Transcription Factor Interactome
Shokri L, et al.
Cell Reports, 27(3), 955-970 (2019)
Noggin4 is a long-range inhibitor of Wnt8 signalling that regulates head development in Xenopus laevis
Eroshkin FM, et al.
Scientific Reports, 6, 23049-23049 (2016)
Global Trade Item Number
| SKU | GTIN |
|---|---|
| P7582-100UG | 04061834395362 |
| P7582-50UG | 04061833266069 |
| P7582-40UG | 04061833266052 |
| P7582-25UG | 04061833266045 |
-
Does the FLAG-BAP fusion protein also contain a his tag?
1 answer-
Helpful?
-

