Skip to Content
Merck
All Photos(1)

Documents

P7457

Sigma-Aldrich

Carboxy-terminal FLAG-BAP Fusion Protein

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.32

form

liquid

Quality Level

mol wt

~49 kDa

shipped in

dry ice

storage temp.

−20°C

General description

Carboxy-terminal FLAG-BAP Fusion Protein is a 466 amino acid C-terminal FLAG fusion protein of E.coli bacterial alkaline phosphatase (BAP).

Application

Carboxy-terminal FLAG-BAP Fusion Protein has been used in the immunoprecipitation of the reporter protein in human embryonic kidney (HEK) cell lysate and as a FLAG-tagged control protein in solid-phase binding assay of spermatogenic immunoglobulin superfamily protein (SgIGSF).
Learn more product details in our FLAG® application portal.

Biochem/physiol Actions

The FLAG sequence comprises of the eight-amino acid sequence AspTyrLysAspAspAspAspLys and is hydrophilic. FLAG fusion proteins are expressed in bacterial, yeast and mammalian cells. FLAG epitope-tagged bacterial alkaline phosphatase is employed in immunoaffinity purification. Alkaline phosphatase based fusion protein have wide clinical applications in immunodetection, enzyme immunoassay and enzyme-linked immunosorbent assay.

Other Notes

Control protein

Physical form

Supplied in 10 mM Tris, 120 mM NaCl, 0.05 mM ZnCl2

Preparation Note

Dilute the ANTI-FLAG M2 antibody solution to 10 mg/ml

Legal Information

FLAG is a registered trademark of Merck KGaA, Darmstadt, Germany
FLAG-BAP is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cloning of a soluble isoform of the SgIGSF adhesion molecule that binds the extracellular domain of the membrane-bound isoform
Koma Y, et al.
Oncogene, 23(33), 5687-5687 (2004)
Vectors for expression and secretion of FLAG epitope-tagged proteins in mammalian cells
Chubet RG and Brizzard BL
Biotechniques, 20(1), 136-141 (1996)
Eldie Berger et al.
Microbial cell factories, 10, 62-62 (2011-08-05)
Through modification of the flagellin type III secretion pathway of Bacillus halodurans heterologous peptides could be secreted into the medium as flagellin fusion monomers. The stability of the secreted monomers was significantly enhanced through gene-targeted inactivation of host cell extracellular
Patricia L Pelczar et al.
Journal of bacteriology, 190(16), 5635-5641 (2008-06-17)
GerD of Bacillus subtilis is a protein essential for normal spore germination with either L-alanine or a mixture of L-asparagine, D-glucose, D-fructose, and potassium ions. GerD's amino acid sequence suggests that it may be a lipoprotein, indicating a likely location
Maddalena de Virgilio et al.
Journal of experimental botany, 59(10), 2815-2829 (2008-06-10)
Protein bodies (PB) are stable polymers naturally formed by certain seed storage proteins within the endoplasmic reticulum (ER). The human immunodeficiency virus negative factor (Nef) protein, a potential antigen for the development of an anti-viral vaccine, is highly unstable when

Articles

Comparison of elution techniques for small-scale protein purification of FLAG® tag proteins using anti-FLAG® M2 magnetic beads.

Protocols

Protocol for immunoprecipitation (IP) of FLAG fusion proteins using M2 monoclonal antibody 4% agarose affinity gels

Related Content

Protein purification techniques, reagents, and protocols for purifying recombinant proteins using methods including, ion-exchange, size-exclusion, and protein affinity chromatography.

Protein purification techniques, reagents, and protocols for purifying recombinant proteins using methods including, ion-exchange, size-exclusion, and protein affinity chromatography.

Protein purification techniques, reagents, and protocols for purifying recombinant proteins using methods including, ion-exchange, size-exclusion, and protein affinity chromatography.

Protein purification techniques, reagents, and protocols for purifying recombinant proteins using methods including, ion-exchange, size-exclusion, and protein affinity chromatography.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service