Skip to Content
Merck

Skip To

P162

Pregnenolone sulfate sodium salt

sigma-1 receptor agonist, powder

Synonym(s):

5-Pregnen-3β-ol-20-one sulfate, 3β-Hydroxy-5-pregnen-20-one 3-sulfate, 3-Hydroxypregn-5-en-20-one sulfate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C21H31NaO5S
Molecular Weight:
418.52
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
MDL number:
Form:
powder
Quality level:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Pregnenolone sulfate sodium salt,

form

powder

Quality Level

mp

192 °C (lit.)

solubility

methanol and water: 1.93 mg/mL, 45% (w/v) aq 2-hydroxypropyl-β-cyclodextrin: 2 mg/mL

SMILES string

[Na].[H][C@@]12CC=C3C[C@H](CC[C@]3(C)[C@@]1([H])CC[C@]4(C)[C@H](CC[C@@]24[H])C(C)=O)OS(O)(=O)=O

InChI

1S/C21H32O5S.Na.H/c1-13(22)17-6-7-18-16-5-4-14-12-15(26-27(23,24)25)8-10-20(14,2)19(16)9-11-21(17,18)3;;/h4,15-19H,5-12H2,1-3H3,(H,23,24,25);;/t15-,16-,17+,18-,19-,20-,21+;;/m0../s1

InChI key

ABFNHVDDMHKJKK-WRHGONFGSA-N

Application

Pregnenolone sulfate sodium salt has been used as sigma-1 receptor (σ1R) agonist. It has also been used in the stimulation and as a ligand for transient receptor potential melastatin-3 (TRPM3).

Biochem/physiol Actions

Pregnenolone sulfate is a neurosteroid and a neuromodulator. It is involved in the stimulation of N-methyl-D-aspartate receptor (NMDA), γ-Aminobutyric acid type A (GABAA ) and α-amino-3-hydroxy-5-methyl-4-isoxazolepropionic acid (AMPA-kainate). It activates transient receptor potential melastatin (TRPM3) which is a calcium-permeable ion channel. It improves schizophrenia and behavioral anomalies by modulating NMDA mediated Ser/Thr kinase - glycogen synthase kinase 3 β (AKT/GSK3β) signaling pathway.

Features and Benefits

This compound is a featured product for Neuroscience research. Click here to discover more featured Neuroscience products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
D5297P8004721301
form

powder

form

powder

form

powder

form

powder

Quality Level

100

Quality Level

200

Quality Level

100

Quality Level

-

solubility

methanol and water: 1.93 mg/mL, 45% (w/v) aq 2-hydroxypropyl-β-cyclodextrin: 2 mg/mL

solubility

methanol: soluble, clear to hazy

solubility

-

solubility

-

mp

192 °C (lit.)

mp

148-149 °C (dec.) (lit.)

mp

-

mp

192 °C (lit.)

Gene Information

human ... GABRA1(2554), GABRA2(2555), GABRA3(2556), GABRA4(2557), GABRA5(2558), GABRA6(2559), GABRB1(2560), GABRB2(2561), GABRB3(2562)

Gene Information

human ... GABRA1(2554), GABRA2(2555), GABRA3(2556), GABRA4(2557), GABRA5(2558), GABRA6(2559), GABRB1(2560), GABRB2(2561), GABRB3(2562)

Gene Information

-

Gene Information

-


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



CREB, AP-1, ternary complex factors and MAP kinases connect transient receptor potential melastatin-3 (TRPM3) channel stimulation with increased c-Fos expression
Rubil S, et al.
British Journal of Pharmacology, 173(2), 305-318 (2016)
Pregnenolone sulfate normalizes schizophrenia-like behaviors in dopamine transporter knockout mice through the AKT/GSK3beta pathway
Wong P, et al.
Translational Psychiatry, 5(3), e528-e528 (2015)
Pregnenolone sulfate as a modulator of synaptic plasticity
Smith CC, et al.
Psychopharmacology, 231(17), 3537-3556 (2014)



Global Trade Item Number

SKUGTIN
P162-25MG04061832083612
P162-100MG04061834355908

Questions

Reviews

No rating value

Active Filters