Skip to Content
Merck
All Photos(3)

Key Documents

O7252

Sigma-Aldrich

Orange G

Dye content, ≥80%, certified by the Biological Stain Commission, powder

Synonym(s):

1-Phenylazo-2-naphthol-6,8-disulfonic acid disodium salt, 7-Hydroxy-8-phenylazo-1,3-naphthalenedisulfonic acid disodium salt, Acid Orange 10, Wool Orange 2G

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H10N2Na2O7S2
CAS Number:
Molecular Weight:
452.37
Colour Index Number:
16230
Beilstein:
4120705
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47
Pricing and availability is not currently available.

Product Name

Orange G, certified by the Biological Stain Commission

grade

certified by the Biological Stain Commission

Quality Level

form

powder

composition

Dye content, ≥80%

technique(s)

microbe id | staining: suitable

color

orange to very dark orange

solubility

water: 1 mg/mL, clear

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Na+].[Na+].Oc1ccc2cc(cc(c2c1\N=N\c3ccccc3)S([O-])(=O)=O)S([O-])(=O)=O

InChI

1S/C16H12N2O7S2.2Na/c19-13-7-6-10-8-12(26(20,21)22)9-14(27(23,24)25)15(10)16(13)18-17-11-4-2-1-3-5-11;;/h1-9,19H,(H,20,21,22)(H,23,24,25);;/q;2*+1/p-2/b18-17+;;

InChI key

HSXUHWZMNJHFRV-QIKYXUGXSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Orange G (C.l. 16230) is the disodium salt of benzeneazo-2-naphthol-6,8-disulfonic acid, C16H10N2O7S2Na2. Orange G, also known as wool orange 2G, is a component in several industry applications including cytology, histology, biology, and industrial staining. It is a component of Mallory′s connective tissue stain.

Application

Orange G is typically used as a background or cytoplasmic stain for various biological samples including a common application within clincial cytology within Papanicolaou varients.

Suitability

Certified for use in Mallory′s connective tissue stain; Wilson-Ezrin method for pituitary acidophils; Kreyberg′s method for keratin and mucus; as a counterstain following Heidenhain′s iron hematoxylin on paraffin sections; Flemming′s triple stain on paraffin sections of onion root.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Integrity of chromatin and replicating DNA in nuclei released from fission yeast by semi-automated grinding in liquid nitrogen.
Givens RM
BMC Research Notes, 4, 499-499 (2011)
G Thennarasu et al.
Environmental science and pollution research international, 19(7), 2755-2765 (2012-02-23)
The photocatalytic degradation of Orange G (OG) dye has been investigated using synthesised nanocrystalline ZnO as a photocatalyst and sunlight as the irradiation source. The formation of ZnO prepared from its precursor was confirmed through FT-IR and powder X-ray diffraction
M Arulkumar et al.
Journal of hazardous materials, 186(1), 827-834 (2011-01-15)
Thespesia populnea is a large tree found in the tropical regions and coastal forests of India. Its pods were used as a raw material for the preparation of activated carbon. The prepared activated carbon was used for the adsorptive removal
Arumugam Sivasamy et al.
Current microbiology, 62(2), 351-357 (2010-07-21)
Biosorption is an eco-friendly and cost-effective method for treating the dye house effluents. Aspergillus niger and Trichoderma sp. were cultivated in bulk and biomasses used as biosorbents for the biosorption of an azo dye Orange G. Batch biosorption studies were
A R Khataee et al.
Journal of hazardous materials, 168(1), 451-457 (2009-03-13)
In order to discuss the effect of chemical structure on photocatalysis efficiency, the photocatalytic degradation of three commercial textile dyes (C.I. Acid Orange 10 (AO10), C.I. Acid Orange 12 (AO12) and C.I. Acid Orange 8 (AO8)) with different structure and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service