Skip to Content
Merck
All Photos(3)

Key Documents

195235

Sigma-Aldrich

Orange II sodium salt

(Certified by the Biological Stain Commission), Dye content ≥85 %

Synonym(s):

4-(2-Hydroxy-1-naphthylazo)benzenesulfonic acid sodium salt, Acid Orange 7, Acid Orange A, Orange II, Tropaeolin 000 No. 2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H11N2NaO4S
CAS Number:
Molecular Weight:
350.32
Colour Index Number:
15510
Beilstein:
3898201
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

product line

BioXtra

form

powder

composition

Dye content, ≥85%

λmax

483 nm

ε (extinction coefficient)

≥17000 at 480-486 nm in water at 0.01 g/L
≥26500 at 225-231 nm in water at 0.01 g/L
≥7500 at 307-313 nm in water at 0.01 g/L

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Na+].Oc1ccc2ccccc2c1\N=N\c3ccc(cc3)S([O-])(=O)=O

InChI

1S/C16H12N2O4S.Na/c19-15-10-5-11-3-1-2-4-14(11)16(15)18-17-12-6-8-13(9-7-12)23(20,21)22;/h1-10,19H,(H,20,21,22);/q;+1/p-1/b18-17+;

InChI key

CQPFMGBJSMSXLP-ZAGWXBKKSA-M

Looking for similar products? Visit Product Comparison Guide

Suitability

Certified for use in the Wilson-Ezrin method for pituitary acidophils, as a counterstain following Heidenhain′s or Weigert′s hematoxylin on paraffin sections, and as a substitute for orange G in Floemming′s triple stain.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Liyuan Kuang et al.
Journal of environmental monitoring : JEM, 13(9), 2496-2501 (2011-08-13)
Mesoporous TiO(2) microspheres were prepared by a hydrothermal reaction and are characterized in this paper. Decoloration and mineralization during photodegradation of Orange II by mesoporous TiO(2) at different pH values, formation of sulfate, relative luminosity to luminous bacteria and recycling
Ruixia Yuan et al.
Journal of hazardous materials, 196, 173-179 (2011-10-05)
Sodium chloride is a common salt used during textile wet processes. Here a dual effect of chloride (i.e. inhibitory and accelerating effect) on azo dye (Acid Orange 7, AO7) degradation in an emerging cobalt/peroxymonosulfate (Co/PMS) advanced oxidation process (AOP) was
Xin Yang et al.
Huan jing ke xue= Huanjing kexue, 32(7), 1960-1966 (2011-09-20)
Persulfate oxidation is a recently emerging advanced oxidation process for the pollution control. Persulfate (PS) degradation of azo dye acid orange 7 (AO7) in an aqueous solution was studied in the presence of suspended granular activated carbon (GAC) at ambient
Mohammad A Behnajady et al.
Journal of nanoscience and nanotechnology, 13(1), 548-553 (2013-05-08)
In this study, monometallic and bimetallic silver and copper doped TiO2 nanoparticles were prepared by sol-gel method. Structural and morphological characterizations of prepared nanoparticles were performed by X-ray diffraction (XRD), transmission electron microscopy (TEM), scanning electron microscopy (SEM), energy dispersive
S S Shinde et al.
Journal of photochemistry and photobiology. B, Biology, 117, 262-268 (2012-11-21)
Ag-doped ZnO thin films with preferred c-axis orientation along (002) have been prepared by spray pyrolysis technique in aqueous medium on to the corning glass substrates. The effect of Ag-doping on to the photoelectrochemical, structural, morphological, optical, luminescence, electrical and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service