Skip to Content
Merck
All Photos(3)

Key Documents

N0636

Sigma-Aldrich

Nicotinamide

BioReagent, suitable for cell culture, suitable for insect cell culture

Synonym(s):

Niacinamide, Nicotinic acid amide, Pyridine-3-carboxylic acid amide, Vitamin B3, Vitamin PP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6N2O
CAS Number:
Molecular Weight:
122.12
Beilstein:
383619
EC Number:
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.75

biological source

synthetic (organic)

product line

BioReagent

Assay

≥98% (HPLC)

form

powder

mol wt

Mw 122.12 g/mol

technique(s)

cell culture | insect: suitable
cell culture | mammalian: suitable

color

white

mp

128-131 °C (lit.)

solubility

water: soluble 50 mg/mL, clear, colorless

SMILES string

NC(=O)c1cccnc1

InChI

1S/C6H6N2O/c7-6(9)5-2-1-3-8-4-5/h1-4H,(H2,7,9)

InChI key

DFPAKSUCGFBDDF-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Nicotinamide (NAM) belongs to vitamin B family and is the amide form of vitamin B3. NAM is present in food and is a constituent of nicotinamide adenine dinucleotide (NAD) and nicotinamide adenine dinucleotide phosphate (NADP). Tryptophan is the precursor for NAM synthesis in liver. NAM controls cell metabolism, redox reactions, mitochondrial functionsand energy generation. It promotes natural killer cells proliferation and survival. NAM confers neuroprotection and promotes neuronal maturation. It has the ability to cross the blood-brain barrier and may provide protection in ischaemic stroke and traumatic brain injury. NAM may elicit therapeutic benefits in Alzheimer′s, Parkinson′s, and Huntington′s diseases.

Application

Nicotinamide has been used as a medium supplement in organoid culture of intestinal stem cells, esophageal squamous cell carcinomas(ESCC), pancreatic cancer organoids and pancreatic progenitor induction.

Biochem/physiol Actions

Nicotinamide is an amide derivative of vitamin B3 and a PARP inhibitor

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

302.0 °F - closed cup

Flash Point(C)

150 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Human Organoids Share Structural and Genetic Features with Primary Pancreatic Adenocarcinoma Tumors
Romero-Calvo I, et al.
Molecular Cancer Research, molcanres-molca0531 (2018)
Rapid prototyping of a multilayer microphysiological system for primary human intestinal epithelial culture
Hosic S, et al.
bioRxiv, 400721-400721 (2018)
Nicotinamide, a form of vitamin B3, promotes expansion of natural killer cells that display increased in vivo survival and cytotoxic activity
Frei GM, et al.
Blood, 118, 4035-4035 (2011)
Three-dimensional organoids reveal therapy resistance of esophageal and oropharyngeal squamous cell carcinoma cells
Kijima T, et al.
Cellular and molecular gastroenterology and hepatology, 7(1), 73-91 (2019)
Nicotinamide: Mechanism of action and indications in dermatology
Bains P, et al.
Indian Journal of Dermatology, Venereology and Leprology, 84(2), 234-237 (2018)

Articles

Human pancreatic cancer organoid biobank (PDAC organoids) with various KRAS mutations to aide in 3D cell culture and cancer research applications.

Human pancreatic cancer organoid biobank (PDAC organoids) with various KRAS mutations to aide in 3D cell culture and cancer research applications.

Human pancreatic cancer organoid biobank (PDAC organoids) with various KRAS mutations to aide in 3D cell culture and cancer research applications.

Human pancreatic cancer organoid biobank (PDAC organoids) with various KRAS mutations to aide in 3D cell culture and cancer research applications.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service