Skip to Content
Merck

Skip To

MSQC9

SILuMAb Infliximab Stable-Isotope Labeled Monoclonal Antibody

recombinant, expressed in CHO cells

Synonym(s):

Mass spectrometry standard, Infliximab, SIL Infliximab, Stable isotope labelled Infliximab

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

NACRES:
NA.41
UNSPSC Code:
12352200
Clone:
-
Species reactivity:
-
Application:
Citations:
1
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


recombinant

expressed in CHO cells

Quality Segment

antibody product type

primary antibodies

assay

≥90% (SDS-PAGE)

packaging

vial of 100 μg

shipped in

wet ice

storage temp.

−20°C

General description

SILuMAb Infliximab Stable-Isotope Labeled Monoclonal Antibody (MSQC9) is a recombinant, stable isotope-labeled, monoclonal antibody which incorporates [13C6, 15N4]-Arginine and [13C6, 15N2]-Lysine. Expressed in CHO cells, it is designed to be used as an internal standard for the quantitative mass spectrometry analysis of Infliximab in human serum. Infliximab is a chimeric monoclonal antibody biologic drug that works against tumor necrosis factor alpha (TNF-a) and is used to treat autoimmune diseases.
Infliximab has been approved for the treatment of Crohn′s disease, ulcerative colitis, psoriasis, psoriatic arthritis, ankylosing spondylitis, and rheumatoid arthritis. SILuMAb Infliximab Stable-Isotope Labeled Monoclonal Antibody is for R&D use only. Not for drug, household, or other uses.

Physical form

Each vial of SILuMab Infliximab Stable-Isotope Labeled Monoclonal Antibody contains the labeled antibody in a lyophilized form containing phosphate buffered saline.

Preparation Note

Produced utilizing enriched media containing stable isotope labeled amino acids are 13C6, 15N4-labeled Arginine and 13C6, 15N2-labeled Lysine.
SILuMab Infliximab Stable-Isotope Labeled Monoclonal Antibody is designed to be used as a internal standard for analysis of Infliximab in human serum.
SILuMab Infliximab recovery is maximized when 0.1% formic acid is used for reconstitution of the lyophilized product.Reconstitution with other solvents may reduce recovery. Do not freeze after reconstitution.

  • Briefly centrifuge the vial at ~10,000 × g to collect the product at the bottom of the vial.
  • Add 500 μL of ultrapure water containing 0.1% formic acid to the vial.
  • Mix the contents by gently inverting the vial a minimum of 5 times.
  • Allow the vial to stand at room temperature for at least 15 minutes and repeat mixing by inversion.

Analysis Note

Quantitative
MRM settings provided (xls)

Legal Information

This product is licensed under U.S. Patent No. 7,396,688 and foreign counterparts from E. I. du Pont de Nemours and Company. The purchase of this product conveys to the buyer the nontransferable right to use the purchased amount of the product for research and development only, including services for a third party for consideration. The buyer cannot sell or otherwise transfer this product, its components or materials made using this product or its components to a third party. Information about licenses for excluded uses is available from: E. I. du Pont de Nemours and Company; Attn: Associate Director, Commercial Development; DuPont Experimental Station E268; 200 Powdermill Rd.; Wilmington, DE 19803; 1-877-881-9787 (voice), 1-302-695-1437 (fax), [email protected].
SILu is a trademark of Sigma-Aldrich Co. LLC

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
MSQC11MSQC3MSQC25
shipped in

wet ice

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

packaging

vial of 100 μg

packaging

vial of 100 μg

packaging

vial of 100 μg (± 10% Lot-specific vial content given on certificate of analysis)

packaging

-

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

assay

≥90% (SDS-PAGE)

assay

≥90% (SDS-PAGE)

assay

≥90% (SDS-PAGE)

assay

≥90% (HPLC)

recombinant

expressed in CHO cells

recombinant

expressed in CHO cells

recombinant

expressed in CHO cells

recombinant

expressed in CHO cells


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

13 - Non Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters