Skip to Content
Merck

Skip To

MSQC3

SILuMAB Stable-Isotope Labeled Universal Monoclonal Antibody Standard human

recombinant, expressed in CHO cells

Synonym(s):

Mass spectrometry standard, IgG, Mass spectrometry standard, Immunoglobulin-G, Stable isotope labelled IgG

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

NACRES:
NA.12
UNSPSC Code:
23201100
Clone:
-
Species reactivity:
-
Application:
Citations:
9
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


recombinant

expressed in CHO cells

Quality Segment

assay

≥90% (SDS-PAGE)

form

solid

packaging

vial of 100 μg (± 10% Lot-specific vial content given on certificate of analysis)

shipped in

wet ice

storage temp.

−20°C

General description

SILuMab is a recombinant, stable isotope-labeled, human monoclonal antibody which incorporates [13C6, 15N4]-Arginine and [13C6, 15N2]-Lysine. Expressed in CHO cells, SILuMab is designed to be used as a universal internal standard for bioanalysis of monoclonal antibodies. Because of overlap with common sequences in the Fc region with candidate antibodies, SILuMab provides universal utility and thus eliminates the need to produce candidate-specific internal standards. SILuMab is an IgG1 antibody with lambda light chain, but contains peptide sequences common to other IgG isotypes.

Application

SILuMAB Stable-Isotope Labeled Universal Monoclonal Antibody Standard human has been used in a comparative study of automated antibody de novo sequencing.

Features and Benefits

Universal Peptide SequenceLocation
DTLMISRHeavy Chain (IgG1, IgG2, IgG3, IgG4)
FNWYVDGVEVHNAKHeavy Chain (IgG1)
VVSVLTVLHQDWLNGKHeavy Chain (IgG1, IgG3, IgG4)
NQVSLTCLVKHeavy Chain (IgG1, IgG2, IgG3, IgG4)
GFYPSDIAVEWESNGQPENNYKHeavy Chain (IgG1, IgG4)
AGVETTTPSKLight Chain (lambda)
YAASSYLSLTPEQWKLight Chain (lambda)
SILuMab has been validated as an internal standard for quantitation of relevant biotherapeutics in a complex biological matrix by MRM-based LC-MS/MS.
  • SILuMab yielded reproducible, linear curves from 0.1 μg/mL to 1000 μg/mL without enrichment or depletion.
  • Good agreement was observed between multiple peptides derived from the same target.
  • Label incorporation was determined to be >98% by mass spectrometry.
  • Sequence coverage was confirmed by peptide mapping.

Physical form

Supplied as a lyophilized powder containing phosphate buffered saline

Preparation Note

Produced utilizing enriched media containing stable isotope labeled amino acids are 13C6, 15N4-labeled arginine and 13C6, 15N2-labeled lysine
SILuMab design is optimized to be used as an internal standard for quantitation of monoclonal antibodies as well as Fc-fusion therapeutics. Because of overlap with the common sequences in the Fc region with candidate antibodies, SILuMab provides univer­sal utility, thus eliminating the need for production of candidate-specific internal standards.
SILuMab recovery is maximized when 0.1% formic acid is used for reconstitution of the lyophilized product. Reconstitution with other solvents may reduce recovery. Do not freeze after reconstitution.
Procedure
  • Briefly centrifuge the vial at ~10,000 x g to collect the product at the bottom of the vial.
  • Add 500 μL of purified water containing 0.1% formic acid to the vial.
  • Mix the contents by gently inverting the vial a minimum of 5 times.
  • Allow the vial to stand at room temperature for a minimum of 15 minutes and repeat mixing by inversion.

Analysis Note

SILuMab Heavy Chain
EVQLVESGGGLVQPGGSLRLSCVASGFTLNNYDMHWVRQGIGKGLEWVSKI
GTAGDRYYAGSVKGRFTISRENAKDSLYLQMNSLRVGDAAVYYCARGAGRW
APLGAFDIWGQGTMVTVSS|ASTKGPSVFPLAPSSKSTSGGTAALGCLVKDYF
PEPVTVSWNSGALTSGVHTFPAVLQSSGLYSLSSVVTVPSSSLGTQTYICNVN
HKPSNTKVDKKVEPKSCDKTHTCPPCPAPELLGGPSVFLFPPKPKDTLMISR
TPEVTCVVVDVSHEDPEVKFNWYVDGVEVHNAKTKPREEQYNSTYRV
VSVLTVLHQDWLNGKEYKCKVSNKALPAPIEKTISKAKGQPREPQVYTLPPS
RDELTKNQVSLTCLVKGFYPSDIAVEWESNGQPENNYKTTPPVLDSDGSFFL
YSKLTVDKSRWQQGNVFSCSVMHEALHNHYTQKSLSLSPG

SILuMab Light Chain
QSALTQPRSVSGSPGQSVTISCTGTSSDIGGYNFVSWYQQHPGKAPKLMIY
DATKRPSGVPDRFSGSKSGNTASLTISGLQAEDEADYYCCSYAGDYTPGV
VFGGGTKLTVL|GQPKAAPSVTLFPPSSEELQANKATLVCLISDFYPGAVTV
AWKADSSPVKAGVETTTPSKQSNNKYAASSYLSLTPEQWKSHRSYSCQ
VTHEGSTVEKTVAPTECS

Target overlap areas are underlined
Qualitative
Intact heavy and light chains (FASTA file)

Quantitative
MRM settings provided (Skyline, xls)
Package size based on protein content determined by A280 using an extinction coefficient (E0.1%) of 1.4

Legal Information

This product is licensed under U.S. Patent No. 7,396,688 and foreign counterparts from E. I. du Pont de Nemours and Company. The purchase of this product conveys to the buyer the nontransferable right to use the purchased amount of the product for research and development only, including services for a third party for consideration. The buyer cannot sell or otherwise transfer this product, its components or materials made using this product or its components to a third party. Information about licenses for excluded uses is available from: E. I. du Pont de Nemours and Company; Attn: Associate Director, Commercial Development; DuPont Experimental Station E268; 200 Powdermill Rd.; Wilmington, DE 19803; 1-877-881-9787 (voice), 1-302-695-1437 (fax), [email protected].
SILu is a trademark of Sigma-Aldrich Co. LLC

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
MSQC11MSQC7MSQC29
storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

form

solid

form

-

form

-

form

solid

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice

packaging

vial of 100 μg (± 10% Lot-specific vial content given on certificate of analysis)

packaging

vial of 100 μg

packaging

vial of 100 μg (± 10% Lot-specific vial content given on certificate of analysis)

packaging

-

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

assay

≥90% (SDS-PAGE)

assay

≥90% (SDS-PAGE)

assay

≥90% (SDS-PAGE)

assay

≥90% (HPLC)


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters