Skip to Content
Merck

Skip To

M8410

Mineral oil

light oil, (neat)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
12352207
NACRES:
NA.75
EC Number:
232-455-8
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

sterility

non-sterile

form

light oil

storage condition

protect from light

refractive index

n20/D 1.467 (lit.)

density

0.84 g/mL at 25 °C (lit.)

format

neat

storage temp.

room temp

InChI key

AEOVEGJBKQQFOP-DDVLFWKVSA-L

Application

Mineral oil has been used as an overlay in microdrop embryo cultures.

Analysis Note

This material is tested as an overlay in microdrop embryo cultures. (All embryo tested materials are tested using mouse embryos.) Typically BCF1 x BDF1 hybrids are used. The oil is primarily used to prevent evaporation of the medium, particiularly when performing embryo manipulations.1 The procedure is as follows: 1-cell (BCF1 X BDF1) embryos were cultured in triplicate microdrops of embryo-tested HTF (Human Tubal Fluid) medium supplemented with 0.4% BSA. For a valid assay, at least 70% of 1-cell stage control embryos must develop to the blastocyst stage within 96 hours. For product release, at least 80% of 1-cell stage test embryos must develop to the blastocyst stage within 96 hours with the tested material.

Disclaimer

This product is not sterile. Mineral oil is difficult to sterilize. It can only be sterilized by careful filtration of the warmed oil. It is assumed antibiotics will be added to the cell culture medium. Otherwise, Prod. No. M5310 should be used, as it is sterile filtered.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
M5904M5310330779
Quality Level

200

Quality Level

200

Quality Level

300

Quality Level

200

form

light oil

form

light oil

form

light oil

form

viscous liquid

density

0.84 g/mL at 25 °C (lit.)

density

0.84 g/mL at 25 °C (lit.)

density

0.82-0.88 g/mL at 25 °C

density

0.833 g/mL at 25 °C

refractive index

n20/D 1.467 (lit.)

refractive index

n20/D 1.467 (lit.)

refractive index

n20/D 1.467 (lit.)

refractive index

n20/D 1.467 (lit.)

storage condition

protect from light

storage condition

-

storage condition

protect from light

storage condition

-

format

neat

format

-

format

neat

format

-


Still not finding the right product?

Explore all of our products under Mineral oil


Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

No data available

flash_point_c

No data available

ppe

Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



TIE: A Method to Electroporate Long DNA Templates into Preimplantation Embryos for CRISPR-Cas9 Gene Editing
Bagheri H, et al.
The CRISPR journal, 1(3), 223-229 (2018)
Minoru Kubo et al.
Nucleic acids research, 47(9), 4539-4553 (2019-03-16)
Next-generation sequencing technologies have made it possible to carry out transcriptome analysis at the single-cell level. Single-cell RNA-sequencing (scRNA-seq) data provide insights into cellular dynamics, including intercellular heterogeneity as well as inter- and intra-cellular fluctuations in gene expression that cannot
Production of a gonadotropin-releasing hormone 2 receptor knockdown (GNRHR2 KD) swine line
Desaulniers AT, et al.
Transgenic research, 26(4), 567-575 (2017)



Global Trade Item Number

SKUGTIN
M8410-1L04061838259844
M8410-500ML04061838259851
M8410-5ML04061834063070
M8410-100ML04061838259837

Questions

1–5 of 5 Questions  
  1. Can M8410 Sigma-Aldrich Mineral oil be used for the deparaffinization of FFPE samples, and is it compatible with downstream histological or molecular analyses like immunohistochemistry or RNA/DNA extraction?

    1 answer
    1. This product has not been validated for deparaffinization of formalin-fixed, paraffin-embedded samples. However, an article in "Applied Immunohistochemistry & Molecular Morphology" (2014 Sep;22(8):623-6) indicated that mineral oil appears to work well for deparaffinizing these samples and for DNA extraction.

      Helpful?

  2. Hello,I´m looking for a mineral oil for sulfur pattern production. It´s difficult to find an oil in which the content of sulfur compounds appears in the COA. Can you inform if there´s a possibility of sulfur contamination in the production of mineral oil?

    1 answer
    1. The manufacturing process for this product is considered proprietary. Currently, there is no product option available that is tested for the presence of sulfur.

      Helpful?

  3. ıs this suitable for preparation of carbon paste electrode?

    1 answer
    1. This material has not been validated in the process of making carbon paste electrodes. However, if the specifications (found below) meet your requirements, then there is no reason that they can't be used in this process.
      https://www.sigmaaldrich.com/specification-sheets/990/164/M8410-BULK.pdf

      Helpful?

  4. Hello, Does this product have an odour? I want to use it for an olfactory memory assay, so needs to be odourless

    1 answer
    1. According to Section 9 of the Safety Data Sheet, this is an odorless chemical.

      Helpful?

  5. What is the composition of the oil? Where does it comes from?

    1 answer
    1. M8410 is a light oil. Mineral oil is a complex mixture of different hydrocarbons and its typical composition is as follows:

      Paraffinic oils, based on n-alkanes, 15-60%
      Naphthenic oils, based on cycloalkanes, 30%-60%
      Aromatic oils, based on aromatic hydrocarbons, 3-30%
      Light and heavy mineral oil differ in terms of composition, viscosity, and density.
      Mineral oil with an alkyl chain carbon number of 15 to 25 is categorized as light oil whereas 25 to 50 as heavy oil.
      Light oil has a density of 0.83–0.86 kg L-1 whereas heavy oil has 0.875–0.905 kg L-1
      Light mineral oil is less occlusive than heavy mineral oil as it contains more cyclic (saturated) molecules

      Additional Links: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/326/294/m8410pis.pdf and a second document: https://www.sigmaaldrich.com/specification-sheets/990/164/M8410-BULK.pdf

      Helpful?

Reviews

No rating value

Active Filters