Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Mineral oil
Synonym(s):
Mineral oil
CAS Number:
8042-47-5
Products
Product Category
solvents (1)
Brand
Sigma-Aldrich (7)
Supelco (2)
Color
Feature
Physical Form
Product Line
Agency
Format
Quality Segment
Application
Package Size
Special Grade
Reference Material (1)
pharmaceutical secondary standard (1)
Technique
Filter Products
Showing 1-11 of 11
Compare
Product Number
Description
Pricing
M8410
Mineral oil,
light oil, (neat)
Expand
View Pricing
M5904
Mineral oil,
BioReagent, Molecular Biology, light oil
Expand
View Pricing
M5310
Mineral oil,
light oil, suitable for mouse embryo cell culture, BioXtra
Expand
View Pricing
M8662
Mineral oil,
PCR Reagent
Expand
View Pricing
PX0045
Mineral oil,
white
Expand
View Pricing
69794
Mineral oil,
BioUltra, Molecular Biology
Expand
View Pricing
330779
Mineral oil,
light
Expand
View Pricing
330760
Mineral oil,
heavy
Expand
View Pricing
M3516
Mineral oil,
suitable for preparation of Nujol mulls for infrared spectroscopy, light oil
Expand
View Pricing
M1180
Mineral oil,
meets USP testing specifications
Expand
View Pricing
PHR2881
Mineral oil,
pharmaceutical secondary standard, reference material
Expand
View Pricing