Skip to Content
Merck

Skip To

M4267

Mefenamic acid

COX inhibitor

Synonym(s):

2-[(2,3-Dimethylphenyl)amino]benzoic acid, N-(2,3-Xylyl)anthranilic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C15H15NO2
CAS Number:
Molecular Weight:
241.29
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12161501
EC Number:
200-513-1
MDL number:
Quality level:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Mefenamic acid,

Quality Level

originator

Shionogi

SMILES string

Cc1cccc(Nc2ccccc2C(O)=O)c1C

InChI

1S/C15H15NO2/c1-10-6-5-9-13(11(10)2)16-14-8-4-3-7-12(14)15(17)18/h3-9,16H,1-2H3,(H,17,18)

InChI key

HYYBABOKPJLUIN-UHFFFAOYSA-N

Gene Information

Application

Mefenamic acid has been used:
  • to test its hepatotoxic effect in the transgenic zebrafish cell line (LiPan)[1]
  • to test its neuroprotective functionality in zebrafish embryos/larvae[2]
  • in the preparation of mucoadhesive microparticles in hydrogel beads

An NSAID. Circumvents MRP-mediated multidrug resistance. Specifically and significantly potentiates the cytotoxicity of anthracyclines as well as teniposide, VP-16 and vincristine.

Biochem/physiol Actions

Mefenamic acid is an analgesic and anti-inflammatory drug. It acts as a cyclooxygenase (COX) enzyme inhibitor. It is hepatoxic and implicated in liver injury.[1] Contrarily, mefenamic acid elicits neuroprotection in in vivo ischemic stroke models by inhibiting cell toxicity induced by glutamate.[2] Mefenamic due its inhibitory effect on prostaglandin synthesis can be used in reducing edema and ache.

Features and Benefits

This compound is a featured product for ADME Tox research. Click here to discover more featured ADME Tox products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the Dopamine and Norepinephrine Metabolism page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by Shionogi. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
A8486SML1066T0202
Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

originator

Shionogi

originator

-

originator

-

originator

AstraZeneca

Gene Information

human ... PTGS1(5742), PTGS2(5743)

Gene Information

-

Gene Information

-

Gene Information

human ... SCN10A(6336), SCN11A(11280), SCN1A(6323), SCN2A(6326), SCN3A(6328), SCN4A(6329), SCN5A(6331), SCN7A(6332), SCN8A(6334), SCN9A(6335)


Still not finding the right product?

Explore all of our products under Mefenamic acid


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Howard Horng et al.
Chemical research in toxicology, 26(3), 465-476 (2013-02-14)
Mefenamic acid, (MFA), a carboxylic acid-containing nonsteroidal anti-inflammatory drug (NSAID), is metabolized into the chemically reactive conjugates MFA-1-O-acyl-glucuronide (MFA-1-O-G) and MFA-S-acyl-CoA (MFA-CoA), which are both implicated in the formation of MFA-S-acyl-glutathione (MFA-GSH) conjugates, protein-adduct formation, and thus the potential toxicity
Taro Kojima et al.
Pharmaceutical research, 29(10), 2777-2791 (2012-01-06)
The stabilization mechanism of a supersaturated solution of mefenamic acid (MFA) from a solid dispersion with EUDRAGIT(®) EPO (EPO) was investigated. The solid dispersions were prepared by cryogenic grinding method. Powder X-ray diffractometry, in vitro dissolution test, in vivo oral
Janesh Gupta et al.
The New England journal of medicine, 368(2), 128-137 (2013-01-11)
Menorrhagia is a common problem, yet evidence to inform decisions about therapy is limited. In a pragmatic, multicenter, randomized trial, we compared the levonorgestrel-releasing intrauterine system (levonorgestrel-IUS) with usual medical treatment in women with menorrhagia who presented to their primary



Global Trade Item Number

SKUGTIN
M4267-500G04061833630785
M4267-50G04061834058908

Questions

Reviews

No rating value

Active Filters