Skip to Content
Merck
All Photos(1)

Key Documents

A8486

Sigma-Aldrich

N-(p-Amylcinnamoyl)anthranilic acid

≥98% (HPLC)

Synonym(s):

2-[[1-oxo-3-(4-Pentylphenyl)-2-propen-1-yl]amino]-benzoic acid, ACA, N-(4-Pentylcinnamoyl)anthranilic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H23NO3
CAS Number:
Molecular Weight:
337.41
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to off-white

solubility

DMSO: ≥20 mg/mL

storage temp.

room temp

SMILES string

CCCCCc1ccc(\C=C\C(=O)Nc2ccccc2C(O)=O)cc1

InChI

1S/C21H23NO3/c1-2-3-4-7-16-10-12-17(13-11-16)14-15-20(23)22-19-9-6-5-8-18(19)21(24)25/h5-6,8-15H,2-4,7H2,1H3,(H,22,23)(H,24,25)/b15-14+

InChI key

GAMRBCZMOOMBSQ-CCEZHUSRSA-N

Related Categories

General description

N-(p-Amylcinnamoyl)anthranilic acid (ACA) functions as a broad spectrum phospholipase A2 inhibitor and blocks transient receptor potential (TRP) channels. It inhibits calcium activated chloride current in cardiac ventricular myocytes. ACA regulates various ion channels.

Application

N-(p-Amylcinnamoyl)anthranilic acid has been used as a transient receptor potential cation channel subfamily M member 2 (TRPM2) inhibitor, in studying its role in regulating the production of reactive oxygen species (ROS).
N-(p-Amylcinnamoyl)anthranilic acid is a broad spectrum PLA2 inhibitor and TRP channel blocker. N-(p-Amylcinnamoyl)anthranilic acid has been used to study the functional expression of TRPM2 channels in dopaminergic SNc neurons.

Biochem/physiol Actions

Broad spectrum phospholipase A2 (PLA2) inhibitor and TRP channel blocker.

Features and Benefits

This compound is featured on the Phospholipase A2 page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bo Pang et al.
Pflugers Archiv : European journal of physiology, 463(2), 309-318 (2011-10-25)
Sustained increase in [Ca(2+)](c) (Δ[Ca(2+)](c)) is a critical early signal from T-cell receptor (TCR/CD3). In general, Ca(2+)-release activated Ca(2+) channels (CRAC) are responsible for the Ca(2+) influx and Δ[Ca(2+)](c) after TCR/CD3 stimulation. However, T cells also express Ca(2+)-permeable nonselective cation
Mathias Gelderblom et al.
Stroke, 45(11), 3395-3402 (2014-09-23)
Brain injury during stroke results in oxidative stress and the release of factors that include extracellular Ca(2+), hydrogen peroxide, adenosine diphosphate ribose, and nicotinic acid adenine dinucleotide phosphate. These alterations of the extracellular milieu change the activity of transient receptor
M Göl et al.
Archives italiennes de biologie, 157(1), 24-36 (2019-05-22)
N-(p-amylcinnamoyl) anthranilic acid (ACA) is a blocker of transient receptor potential melastatin-2 (TRPM2) which is a non-selective, Ca2+-permeable and oxidative stress sensor cation channel. Intracerebroventricular (ICV) streptozotocin (STZ) induction successfully generates spatial memory deficits in rats. The purpose of this
Ramazan Bal et al.
European journal of pharmacology, 882, 173163-173163 (2020-06-03)
Oxidative stress-induced Ca2+ permeable transient receptor potential melastatin 2 (TRPM2) channels are expressed at high levels in the brain, appear to link neuronal excitability to cellular metabolism, and are involved in the pathogenesis of neurodegenerative disorders. We aimed to study

Articles

Phospholipase A2 (PLA2) enzymes hydrolyze glycerophospholipids, yielding arachidonic acid; inhibition targets eicosanoid biosynthesis.

Phospholipase A2 (PLA2) enzymes hydrolyze glycerophospholipids, yielding arachidonic acid; inhibition targets eicosanoid biosynthesis.

Phospholipase A2 (PLA2) enzymes hydrolyze glycerophospholipids, yielding arachidonic acid; inhibition targets eicosanoid biosynthesis.

Phospholipase A2 (PLA2) enzymes hydrolyze glycerophospholipids, yielding arachidonic acid; inhibition targets eicosanoid biosynthesis.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service