Skip to Content
Merck

Skip To

M2393

Magnesium chloride hexahydrate

BioReagent, suitable for cell culture, suitable for insect cell culture

Synonym(s):

MgCl2 hexahydrate, MgCl2.6H2O

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Linear Formula:
MgCl2 · 6H2O
CAS Number:
Molecular Weight:
203.30
NACRES:
NA.71
PubChem Substance ID:
UNSPSC Code:
12352302
EC Number:
232-094-6
MDL number:
Form:
powder or crystals
Solubility:
H2O: soluble 500 mg/mL, clear, colorless
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


product line

BioReagent

Quality Segment

form

powder or crystals

technique(s)

cell culture | insect: suitable, cell culture | mammalian: suitable

solubility

H2O: soluble 500 mg/mL, clear, colorless

SMILES string

O.O.O.O.O.O.Cl[Mg]Cl

InChI

1S/2ClH.Mg.6H2O/h2*1H;;6*1H2/q;;+2;;;;;;/p-2

InChI key

DHRRIBDTHFBPNG-UHFFFAOYSA-L

Application

Magnesium chloride hexahydrate (MgCl2·6H2O) has been used:
  • to prepare electrochemical substrate solution for surface treatment and immunoassay preparation of monoclonal anti-creatine kinase (CK-MB) antibody
  • as a component of casamino acids medium (Hv-Ca) to culture Haloferax volcanii strains
  • to prepare hydrated-salt poly(vinylidenefluoride)-hexafluoropropylene P(VDF-HFP)/MgCl2·6H2O nanocomposite film to study its phase transformation, surface morphology and dielectric property

Biochem/physiol Actions

Magnesium chloride is commonly used as a reagent in chemistry and molecular biology as a source of magnesium ions. Magnesium exhibits several biological roles in enzymology, cell membrane and wall structural integrity, muscle cell physiology, and nucleic acid structure. It functions as a co-factor in various enzymes, like deoxyribonuclease (DNAse), the restriction enzymes EcoRI and EcoRV, and Ribonuclease H. Magnesium stabilizes polymeric nucleic acids including ribozymes and transfer RNA.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
M927263068442615-M
form

powder or crystals

form

powder or crystals

form

solid

form

crystalline solid

solubility

H2O: soluble 500 mg/mL, clear, colorless

solubility

-

solubility

H2O: 1 M at 20 °C, clear, colorless

solubility

water: 1.5 g/mL

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

100

technique(s)

cell culture | insect: suitable

technique(s)

-

technique(s)

-

technique(s)

-

product line

BioReagent

product line

-

product line

BioUltra

product line

-

divalent cation, cell attachment, cell adherence, adenovirus infections

-

-

-


Still not finding the right product?

Explore all of our products under Magnesium chloride hexahydrate


Storage Class

13 - Non Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content

Product Information Sheet


Goli Ardestani et al.
Cell calcium, 87, 102181-102181 (2020-02-26)
Prior to maturation, mouse oocytes are arrested at the germinal vesicle (GV) stage during which they experience constitutive calcium (Ca2+) influx and spontaneous Ca2+ oscillations. The oscillations cease during maturation but Ca2+ influx continues, as the oocytes' internal stores attain
Vladimir Riazanski et al.
iScience, 23(11), 101759-101759 (2020-12-01)
The relative contribution of the two phagosomal catabolic processes, oxidative and metabolic, was assessed in the killing of Pseudomonas aeruginosa in phagosomes of alveolar macrophages (AMs) from wild-type (p47-phox +/+ ) or NOX-defective (p47-phox -/- ) mice. Free radical release
Huiwen Hao et al.
EMBO reports, 21(3), e48385-e48385 (2020-01-28)
Microtubules derived from the Golgi (Golgi MTs) have been implicated to play critical roles in persistent cell migration, but the underlying mechanisms remain elusive, partially due to the lack of direct observation of Golgi MT-dependent vesicular trafficking. Here, using super-resolution



Global Trade Item Number

SKUGTIN
M2393-500G04061835546657
M2393-100G04061835546640

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters