Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Magnesium chloride hexahydrate
Linear Formula:
MgCl
2
· 6H
2
O
CAS Number:
7791-18-6
Molecular Weight:
203.30
EC Number:
232-094-6
Products
Product Category
salts (15)
Brand
Millipore (1)
SAFC (3)
Sigma-Aldrich (2)
Supelco (1)
Feature
Formula Weight
pH Value
Physical Form
Product Line
Purity
Agency
Quality Segment
Application
Special Grade
ACS reagent (1)
Puriss (1)
Technique
Filter Products
Showing 1-17 of 17
Compare
Product Number
Description
Pricing
M9272
Magnesium chloride hexahydrate,
ACS reagent, 99.0-102.0%
Expand
View Pricing
M0250
Magnesium chloride hexahydrate,
ReagentPlus
®
, ≥99.0%
Expand
View Pricing
1.05833
Magnesium chloride hexahydrate,
for analysis EMSURE
®
ACS,ISO,Reag. Ph Eur
Expand
View Pricing
13152
Magnesium chloride hexahydrate,
puriss., meets analytical specification of Ph. Eur., BP, FCC, E511, 99-101%, ≤0.0001% Al
Expand
View Pricing
M2393
Magnesium chloride hexahydrate,
BioReagent, suitable for cell culture, suitable for insect cell culture
Expand
View Pricing
1.05832
Magnesium chloride hexahydrate,
crystalline, EMPROVE
®
EXPERT, Ph. Eur., BP, USP, JPC, FCC, E 511
Expand
View Pricing
255777
Magnesium chloride hexahydrate,
99.995% trace metals basis
Expand
View Pricing
M2670
Magnesium chloride hexahydrate,
BioXtra, ≥99.0%
Expand
View Pricing
442615-M
Magnesium chloride hexahydrate,
ACS Reagent Grade.
Expand
View Pricing
1.01872
Magnesium chloride hexahydrate,
crystalline, Ph. Eur., JPC, USP, FCC, E 511
Expand
View Pricing
930989
Magnesium chloride hexahydrate,
99.999% (trace metals basis)
Expand
View Pricing
1.72571
Magnesium chloride hexahydrate,
extra pure, for table water, FCC, E 511
Expand
View Pricing
930970
Magnesium chloride hexahydrate,
99.9% trace metals basis
Expand
View Pricing
63068
Magnesium chloride hexahydrate,
BioUltra, Molecular Biology, ≥99.0% (KT)
Expand
View Pricing
M7304
Magnesium chloride hexahydrate,
meets USP testing specifications
Expand
View Pricing
63064
Magnesium chloride hexahydrate,
BioUltra, ≥99.0% (KT)
Expand
View Pricing
63033
Magnesium chloride hexahydrate,
tested according to Ph. Eur.
Expand
View Pricing