Skip to Content
Merck

Skip To

I6279

ProteoMass Insulin MALDI-MS Standard

vial of 10 nmol, monoisotopic mol wt 5,729.6087 Da

Synonym(s):

Insulin from bovine pancreas

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C254H377N65O75S6
CAS Number:
Molecular Weight:
5733.49
UNSPSC Code:
12352209
NACRES:
NA.24
EC Number:
234-291-2
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


mol wt

monoisotopic mol wt 5,729.6087 Da

Quality Segment

packaging

vial of 10 nmol

InChI key

IXIBAKNTJSCKJM-BUBXBXGNSA-N

Application

Peptide calibration standard for MALDI-MS instruments.

Biochem/physiol Actions

Two-chain polypeptide hormone produced by the β-cells of pancreatic islets. Its molecular weight is ~5800 Da. The α and β chains are joined by two interchain disulfide bonds. The α chain contains an intrachain disulfide bond. Insulin regulates the cellular uptake, utilization, and storage of glucose, amino acids, and fatty acids and inhibits the breakdown of glycogen, protein, and fat.

Other Notes

InChI: 1S/C254H377N65O75S6/c1-29-131(24)205(312-192(334)103-256)250(389)317-204(130(22)23)246(385)287-158(74-81-199(344)345)216(355)281-155(70-77-188(260)330)220(359)307-183-116-399-400-117-184-242(381)305-177(110-321)238(377)293-161(87-122(6)7)223(362)294-167(94-139-52-60-145(324)61-53-139)226(365)282-153(68-75-186(258)328)217(356)289-160(86-121(4)5)221(360)284-157(73-80-198(342)343)219(358)301-173(100-189(261)331)233(372)297-169(96-141-56-64-147(326)65-57-141)229(368)308-182(241(380)303-175(253(393)394)102-191(263)333)115-398-396-113-180(212(351)270-106-193(335)277-152(71-78-196(338)339)215(354)280-150(50-41-83-268-254(264)265)210(349)269-107-194(336)278-165(92-137-45-35-31-36-46-137)225(364)296-166(93-138-47-37-32-38-48-138)228(367)298-170(97-142-58-66-148(327)67-59-142)236(375)318-206(135(28)323)251(390)319-84-42-51-185(319)244(383)285-151(49-39-40-82-255)213(352)276-134(27)252(391)392)310-248(387)202(128(18)19)315-234(373)163(89-124(10)11)291-227(366)168(95-140-54-62-146(325)63-55-140)295-222(361)159(85-120(2)3)288-207(346)132(25)274-214(353)156(72-79-197(340)341)286-245(384)201(127(16)17)314-235(374)164(90-125(12)13)292-231(370)172(99-144-105-267-119-273-144)300-237(376)176(109-320)279-195(337)108-271-211(350)179(112-395-397-114-181(309-243(183)382)240(379)275-133(26)208(347)304-178(111-322)239(378)316-203(129(20)21)249(388)311-184)306-224(363)162(88-123(8)9)290-230(369)171(98-143-104-266-118-272-143)299-218(357)154(69-76-187(259)329)283-232(371)174(101-190(262)332)302-247(386)200(126(14)15)313-209(348)149(257)91-136-43-33-30-34-44-136/h30-38,43-48,52-67,104-105,118-135,149-185,200-206,320-327H,29,39-42,49-51,68-103,106-117,255-257H2,1-28H3,(H2,258,328)(H2,259,329)(H2,260,330)(H2,261,331)(H2,262,332)(H2,263,333)(H,266,272)(H,267,273)(H,269,349)(H,270,351)(H,271,350)(H,274,353)(H,275,379)(H,276,352)(H,277,335)(H,278,336)(H,279,337)(H,280,354)(H,281,355)(H,282,365)(H,283,371)(H,284,360)(H,285,383)(H,286,384)(H,287,385)(H,288,346)(H,289,356)(H,290,369)(H,291,366)(H,292,370)(H,293,377)(H,294,362)(H,295,361)(H,296,364)(H,297,372)(H,298,367)(H,299,357)(H,300,376)(H,301,358)(H,302,386)(H,303,380)(H,304,347)(H,305,381)(H,306,363)(H,307,359)(H,308,368)(H,309,382)(H,310,387)(H,311,388)(H,312,334)(H,313,348)(H,314,374)(H,315,373)(H,316,378)(H,317,389)(H,318,375)(H,338,339)(H,340,341)(H,342,343)(H,344,345)(H,391,392)(H,393,394)(H4,264,265,268)/t131-,132-,133-,134-,135+,149-,150-,151-,152-,153-,154-,155-,156-,157-,158-,159-,160-,161-,162-,163-,164-,165-,166-,167-,168-,169-,170-,171-,172-,173-,174-,175-,176-,177-,178-,179-,180-,181-,182-,183-,184-,185-,200-,201-,202-,203-,204-,205-,206-/m0/s1

Legal Information

ProteoMass is a trademark of Sigma-Aldrich Co. LLC

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
I5500I-035I0516
Quality Level

100

Quality Level

200

Quality Level

300

Quality Level

400

packaging

vial of 10 nmol

packaging

-

packaging

ampule of 0.5 mL

packaging

-

mol wt

monoisotopic mol wt 5,729.6087 Da

mol wt

-

mol wt

-

mol wt

-


Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Yasunari Iguchi et al.
Rapid communications in mass spectrometry : RCM, 31(21), 1845-1850 (2017-08-30)
Continuous flow ionization methods using infrared (IR) lasers have several favorable characteristics, including ionization without any additional matrices and tolerance to contaminants such as detergents and buffer salts. However, poor sensitivity due to low ion-transfer efficiency from the sample plate
Fangqiao Wei et al.
Clinica chimica acta; international journal of clinical chemistry, 509, 101-107 (2020-06-13)
Salivary peptidome profiling analysis has advantages of simplicity and non-invasiveness and great potentiality for screening, monitoring or primary diagnosis of diseases, but may be subjected to change against interferences like diet. We conducted a 5-day study to investigate the influence
Chang Tian et al.
PloS one, 12(8), e0182712-e0182712 (2017-08-16)
This study aimed to identify differences of peptide profiles in stimulated whole saliva among children with and without occurrence of new carious lesions, and to provide a simple way for early diagnosis and prevention of the relapse of severe early



Global Trade Item Number

SKUGTIN
I6279-5X1VL04061833296455
I6279-1VL04061838057099

Questions

Reviews

No rating value

Active Filters