Skip to Content
Merck

Skip To

I4131

IgG from rat serum

reagent grade, ≥95% (SDS-PAGE), essentially salt-free, lyophilized powder

Synonym(s):

Rat IgG

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

UNSPSC Code:
12352203
NACRES:
NA.46
MDL number:
Conjugate:
unconjugated
Species reactivity:
-
Application:
Citations:
88
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


conjugate

unconjugated

Quality Level

grade

reagent grade

assay

≥95% (SDS-PAGE)

form

essentially salt-free, lyophilized powder

storage temp.

2-8°C

target post-translational modification

unmodified

General description

IgG antibody subtype is the most abundant serum immunoglobulins of the immune system. It is secreted by B cells and is found in blood and extracellular fluids, Immunoglobins have two heavy (H) and two light (L) chains, linked by disulphide linkages. Each light chain comprises one variable N-terminal region and a constant C-terminal region. Heavy chain has one variable N-terminal region and three or four constant (CH1-CH4) C-terminal region. The four classes of IgG can be IgG1, IgG2, IgG3, and IgG4.[1] IgG antibody provides protection from infections caused by bacteria, fungi and viruses. Maternal IgG is transferred to fetus through the placenta that is vital for immune defence of the neonate against infections. Rat IgG is purified from pooled normal rat serum by fractionation and ion-exchange chromatography.

Application

IgG from rat serum has been used:
  • in immunolabeling
  • in flow cytometry
  • in enzyme linked immunosorbent assay[2]
  • as a control in neutrophil depletion assay
Purified rat IgG may be used as a reference antigen, standard, blocking agent, or coating protein in a variety of immunoassays including ELISA, dot immunobinding, Western immunoblotting, immunodiffusion, and immunoelectrophoresis. Other applications include starting materials for the preparation of immunogens and solid phase immunoadsorbents. Rat IgG was used as isotype control in flow cytometry[3][4] and injected into mice to induce tolerance against rat IgG.

Biochem/physiol Actions

IgG antibody have enormous therapeutic potential and its fragment crystallizable (Fc) region contributes for the development of therapeutic antibody.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
I8015I5256I5381
conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

storage temp.

2-8°C

storage temp.

−20°C

storage temp.

2-8°C

storage temp.

2-8°C

form

essentially salt-free, lyophilized powder

form

buffered aqueous solution

form

essentially salt-free, lyophilized powder

form

lyophilized powder

grade

reagent grade

grade

technical grade

grade

reagent grade

grade

reagent grade

target post-translational modification

unmodified

target post-translational modification

-

target post-translational modification

unmodified

target post-translational modification

unmodified

assay

≥95% (SDS-PAGE)

assay

≥80% (SDS-PAGE)

assay

≥95% (SDS-PAGE)

assay

≥95% (SDS-PAGE)


Still not finding the right product?

Explore all of our products under IgG from rat serum


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content


Effect of interleukin-6 receptor blockage on renal injury in apolipoprotein E-deficient mice
Tomiyama-Hanayama M et al
American Journal of Physiology: Renal Physiology, 297, F679-F684 (2009)
Catharina D Commerford et al.
Cell reports, 25(13), 3554-3563 (2018-12-28)
Enlargement of the lymphatic vascular network in tumor-draining lymph nodes (LNs) often precedes LN metastasis, likely providing a lymphovascular niche for tumor cells. We investigated morphological and molecular changes associated with the lymphatic remodeling process, using the 4T1 breast cancer
D Hildeman et al.
Journal of virology, 71(12), 9672-9678 (1997-11-26)
Lymphocytic choriomeningitis virus (LCMV) infection of normal mice results in a fatal immunopathologic meningitis mediated by CD8+ cytotoxic T lymphocytes (CTL). We have previously shown that female beta2-microglobulin-deficient (beta2m-/-) mice, which are also deficient in CD8+ T cells, are susceptible



Global Trade Item Number

SKUGTIN
I4131-50MG04061833856635
I4131-10MG04061833856628
I4131-100MG04061833856611

Questions

  1. For Rat IgG product 10MG, I want to resuspend in 50mL PBS to have a final concentration of 0.2mg/mL. Can you let me know the storage after reconstitution (temperature and shelf life please)? Also, do you recommend to aliquot in single use? Thank you!

    1 answer
    1. Upon reconstitution, it is ideal to store in single-use aliquots at –20 °C, as repeated freezing and thawing may compromise stability. The stability upon reconstitution has not been determined.

      Helpful?

Reviews

No rating value

Active Filters