Skip to Content
Merck

Skip To

I5256

IgG from goat serum

reagent grade, ≥95% (SDS-PAGE), essentially salt-free, lyophilized powder

Synonym(s):

Goat IgG

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

UNSPSC Code:
12352203
NACRES:
NA.46
MDL number:
Conjugate:
unconjugated
Species reactivity:
-
Application:
Citations:
64
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


conjugate

unconjugated

Quality Level

grade

reagent grade

assay

≥95% (SDS-PAGE)

form

essentially salt-free, lyophilized powder

storage temp.

2-8°C

target post-translational modification

unmodified

General description

IgG antibody subtype is the most abundant serum immunoglobulins of the immune system. It is secreted by B cells and is found in blood and extracellular fluids and provides protection from infections caused by bacteria, fungi and viruses. Maternal IgG is transferred to fetus through the placenta that is vital for immune defence of the neonate against infections[1]
Goat IgG is purified from normal goat serum by fractionation and ion-exchange chromatography.

Application

Purified IgG may be used as a reference antigen, standard, blocking agent or coating protein in a variety of immunoassays including ELISA, dot immunobinding, Western immunoblotting, immunodiffusion and immunoelectrophoresis. Other applications include starting materials for the preparation of immunogens and solid-phase immunoadsorbents. Goat IgG was used as negative control in immunohistochemical analysis of human heart tissue samples[2] and in preparation of egg-white avidin samples for atomic force microscopy.[3] Goat IgG was used as isotype control antibody in immunohistochemical analysis of tissue sections of human tonsils, colon, small intestine, ileus and spleen.[4]

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
I8015I9140I5006
conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

storage temp.

2-8°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

2-8°C

form

essentially salt-free, lyophilized powder

form

buffered aqueous solution

form

buffered aqueous solution

form

essentially salt-free, lyophilized powder

target post-translational modification

unmodified

target post-translational modification

-

target post-translational modification

-

target post-translational modification

-

assay

≥95% (SDS-PAGE)

assay

≥80% (SDS-PAGE)

assay

≥80% (SDS-PAGE)

assay

≥95% (SDS-PAGE)

grade

reagent grade

grade

technical grade

grade

technical grade

grade

reagent grade


Still not finding the right product?

Explore all of our products under IgG from goat serum


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content


Virginie Gridelet et al.
Reproduction, fertility, and development, 25(3), 511-522 (2013-03-08)
The present study investigated the first interaction that occurs between the blastocyst and endometrium during implantation. Given the ethical objections to studying implantation in humans, a mouse model was used to study the dialogue between luteinising hormone (LH) and luteinising
Mark A Birrell et al.
Journal of immunology (Baltimore, Md. : 1950), 181(6), 4265-4271 (2008-09-05)
The liver X receptors (LXRalpha/beta) are orphan nuclear receptors that are expressed in a large number of cell types and have been shown to have anti-inflammatory properties. Nuclear receptors have previously proved to be amenable targets for small molecular mass
Marlene Brandes et al.
Blood, 102(10), 3693-3701 (2003-07-26)
gamma delta T cells are inadequately defined both in terms of their migration potential and contribution to antimicrobial immunity. Here, we have examined the migration profile of human blood gamma delta T cells and related cell lines and correlated these



Global Trade Item Number

SKUGTIN
I5256-50MG04061833262610
I5256-100MG04061833857199
I5256-10MG04061833262603

Questions

Reviews

No rating value

Active Filters