Skip to Content
Merck

Skip To

FLASC

Adenosine 5′-triphosphate (ATP) bioluminescent somatic cell assay kit

for cellular ATP determination

Synonym(s):

ATP bioluminescence assay kit

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

UNSPSC Code:
12352202
NACRES:
NA.26
eCl@ss:
32160414
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Level

storage temp.

−20°C

General description

Adenosine 5′-triphosphate (ATP) bioluminescent somatic cell assay kit may be employed for the bioluminescent determination of adenosine 5′-triphosphate (ATP) released from a suspension of viable somatic cells. Estimates of cell concentrations may be calculated if it is assumed that the ATP content per viable cell remains constant. The number of viable somatic cells are selectively counted because, as a cell dies, its ATP is rapidly degraded.

Application

The Adenosine 5′-triphosphate (ATP) bioluminescent somatic cell assay kit has been used to determine accurately the level of ATP present in neutrophils isolated from the blood of over 100 human test patients. [1]

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
FLAACS001211699695001
storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

-


Kit Components Only

Product No.
Description

  • FL-AAB Dilution buffer 1 mL/vial

  • FL-AAM Assay mix 1 mL/vial

  • FL-AAS ATP standard 1 mL/vial

  • FL-SAR Somatic cell releasing reagent 1 mL/vial

pictograms

CorrosionEnvironment

signalword

Danger

hcodes

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1

wgk

WGK 3

Storage Class

11 - Combustible Solids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content


Qicheng Ni et al.
Nature communications, 8, 15755-15755 (2017-06-10)
Diabetes is associated with beta cell mass loss and islet dysfunctions. mTORC1 regulates beta cell survival, proliferation and function in physiological and pathological conditions, such as pregnancy and pancreatectomy. Here we show that deletion of Raptor, which is an essential
Lirui Wang et al.
Reproductive toxicology (Elmsford, N.Y.), 75, 23-32 (2017-11-22)
Cadmium (Cd) has been reported to inhibit mouse sperm motility by inducing the tyrosine phosphorylation of dihydrolipoamide dehydrogenase (DLD). This study aimed to assess the potential effects of vitamin C (Vc) on ameliorating Cd-induced tyrosine phosphorylation of DLD and the
Sarah Myhill et al.
International journal of clinical and experimental medicine, 2(1), 1-16 (2009-05-14)
This study aims to improve the health of patients suffering from chronic fatigue syndrome (CFS) by interventions based on the biochemistry of the illness, specifically the function of mitochondria in producing ATP (adenosine triphosphate), the energy currency for all body



Global Trade Item Number

SKUGTIN
FLASC-1KT04061833622919

Questions

1–5 of 5 Questions  
  1. Is any protocol available for this product? What is the working concentration of each solution? Which reagent should be used to prepare dilutions?

    1 answer
    1. Please see the link below to review the product technical bulletin. This document contains the preparation, dilution, and working concentrations of the reagents used: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/313/399/flascbul.pdf

      Helpful?

  2. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  3. What is the optimal dilution for tissue extracts when using Adenosine 5´-triphosphate (ATP) bioluminescent somatic cell assay kit, Product FLASC?

    1 answer
    1. Sigma-Aldrich has not tested this product with tissue extracts. You will need to perform trials to determine optimal dilution of the sample.

      Helpful?

  4. Can Adenosine 5´-triphosphate (ATP) bioluminescent somatic cell assay kit, Product FLASC, be used for adherent cells?

    1 answer
    1. We do not have a separate protocol for adherent cells. However, the somatic cell lysis reagent should be able to lyse cells that are adherent.  The reagent contains enough detergent that the cells will be lysed. Once cells are lysed, it is best to assay the sample immediately.

      Helpful?

  5. Can lysates be stored in the Somatic Cell ATP Releasing Reagent of Adenosine 5´-triphosphate (ATP) bioluminescent somatic cell assay kit, Product FLASC?

    1 answer
    1. In general, no. The working Somatic Cell ATP Releasing Reagent is intended for immediate use, and not for storing ATP in.  We have no specific information as to how long ATP will be stable in it.

      Helpful?

Reviews

No rating value

Active Filters