Skip to Content
Merck

CS0012

ATP Bioluminescence Kit

Sufficient for 200 tests

Synonym(s):

ATP Assay Kit, ATP Detection Kit

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.83
UNSPSC Code:
12161503
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

shipped in

dry ice

storage temp.

−20°C

General description

Adenosine triphosphate (ATP) is a key molecule that stores energy within cells, and is considered as the universal energy currency for all living organisms. ATP has been used as a marker of viability, as living cells have high ATP content whereas dead cells lose their ability to produce ATP. ATP levels are therefore proportional to the amount of living cells in a sample.

The ATP Bioluminescence Kit provides a simple and quick assay to quantify ATP, and also to measure cell viability based on ATP content. This assay is less prone to artifacts than other viability assay methods. Intracellular ATP is released by permeabilization solution and measured using a bioluminescence assay, owing to the ability of luciferase to produce light in the presence of its substrate D-luciferin and ATP.

The kit has the following linear detection range:
  • 0 - 60,000 cells (tested on TF-1 cells)
  • 1nM-10μM ATP

Application

Quantifing ATP within a sample
Measure cell viability based on ATP content

Features and Benefits

Our economical ATP Bioluminescence Kit also saves your time and efforts:
  • Highly sensitive, simple and high-throughput assay for measuring ATP in biological samples
  • User friendly: No need to weigh or calculate needed amounts of reagents on your own
  • Supportive calculator (Click here to download a calculator excel file): will calculate the reagents needed according to your experimental needs, and analyses your results based on your experimental data!
  • Quick instruction bench card - to assure your experimental success
  • More experiments in one kit - contains sufficient reagents for 200 tests


Storage Class

10 - Combustible liquids

wgk

WGK 3



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library