Select a Size
| Pack Size | SKU | Availability | Price |
|---|
About This Item
biological source
goat
Quality Level
conjugate
FITC conjugate
antibody form
affinity isolated antibody
antibody product type
secondary antibodies
clone
polyclonal
form
buffered aqueous solution
contains
≥15 mM sodium azide as preservative
storage condition
protect from light
technique(s)
direct immunofluorescence: 1:160, immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:160
storage temp.
−20°C
target post-translational modification
unmodified
General description
Immunogen
Application
Biochem/physiol Actions
Physical form
Preparation Note
Other Notes
Disclaimer
1 of 1
This Item | |||
|---|---|---|---|
| biological source goat | biological source goat | biological source goat | biological source rabbit |
| antibody form affinity isolated antibody | antibody form affinity isolated antibody | antibody form affinity isolated antibody | antibody form affinity isolated antibody |
| technique(s) direct immunofluorescence: 1:160, immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:160 | technique(s) indirect immunofluorescence: 1:80 | technique(s) direct immunofluorescence: 1:400, immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:400 | technique(s) direct immunofluorescence: 1:400, immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:400 |
| conjugate FITC conjugate | conjugate FITC conjugate | conjugate TRITC conjugate | conjugate FITC conjugate |
| form buffered aqueous solution | form buffered aqueous solution | form buffered aqueous solution | form buffered aqueous solution |
| clone polyclonal | clone polyclonal | clone polyclonal | clone polyclonal |
Still not finding the right product?
Explore all of our products under Anti-Rabbit IgG (whole molecule)–FITC antibody produced in goat
— or —
Try our Product Selector Tool to narrow your options
Storage Class
12 - Non Combustible Liquids
wgk
nwg
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
Instructions
Global Trade Item Number
| SKU | GTIN |
|---|---|
| F9887-1ML | 04061833622155 |
| F9887-2ML | 04061833622216 |
| F9887-.5ML | 04061833622148 |



