Skip to Content
Merck
All Photos(2)

Key Documents

F7367

Sigma-Aldrich

Anti-Goat IgG (whole molecule)–FITC antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Rabbit Anti-Goat IgG (whole molecule)–Fluorescein isothiocyanate

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

rabbit

Quality Level

conjugate

FITC conjugate

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous solution

storage condition

protect from light

technique(s)

direct immunofluorescence: 1:400
immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:400

storage temp.

−20°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

General description

IgG antibodies are smallest and are found in all body fluids and capable of fighting bacterial and viral infection. It is the only type of antibody able to cross the placenta and immunise the foetus. The anti-goat IgG (whole molecule)-FITC antibody is useful in indirect immunofluorescence studies of immunised goat sera infected Trichostrongylus colubriformis (nematode). Antigens can be measured by quantitative immunofluorescence using image analysis, in tissue sections stained with FITC labelled rabbit anti goat IgG. Anti-goat IgG antibody reacts specifically with all goat immunoglobulins.
IgG is a major class of immunoglobulin. Goat IgG has two subclasses- IgG1 and IgG2.
Immunoglobulin G (IgG) is found in blood and has four polypeptide chains. It contains two identical 25 kDa κ or λ light (L) chains and two identical 50 kDa γ heavy (H) chains. The light and heavy chains are interconnected by disulfide bonds. It is the most important type of glycoprotein antibody present in human serum, secreted by the B cells. IgG exists in four isotypes: IgG1, IgG2, IgG3, and IgG4. IgG has the longest serum half-life compared to other immunoglobulins.

Immunogen

IgG purified from Goat

Application

Anti-Goat IgG (whole molecule)−FITC antibody has been used:
  • in double labeling for laminin and α-fetoprotein (AFP) at a dilution of 1:400
  • in immunohistochemistry at a dilution of 1:50
  • in immunocytochemistry
  • in immunochemical jagged-1 analyses
  • in immunofluorescence at a dilution of 1:200
Anti-goat IgG antibody can be used in antisera localization for neurofilament antigens obtained in rat peripheral nerve and can be done in tissues of rat and human peripheral and central nervous system by the aid of indirect immunofluorescence.

Biochem/physiol Actions

Immunoglobulin G (IgG) participates in hypersensitivity type II and type III.
Immunoglobulin G (IgG) stimulates the classical pathway of the complement system. Maternal IgG is transferred to the fetus through the placenta. This is vital for the immune defense of the neonate against infections. IgG regulates antibody-dependent cell-mediated cytotoxicity (ADCC) and might be associated with allergy. It nullifies virus particles and toxins. Digestion of IgG by papain produces fragment antigen-binding (Fab). Pepsin digestion of IgG generates crystallizable (fc) fragment and a single dimeric F(ab)2. IgG levels in severe acute respiratory syndrome coronavirus (SARS-CoV)-2 infected patients may be used as a possible tool to assess the severity and prognosis of coronavirus disease 2019 (COVID-19).

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2-8 °C for up to one month. For extended storage, the solution may be frozen in working aliquots. Repeated freezing and thawing is not recommended. Storage in "frost-free" freezers not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Origin and structural evolution of the early proliferating oval cells in rat liver
Paku S, et al.
The American Journal of Pathology, 158(4), 1313-1323 (2001)
The effect of topical amiloride eye drops on tear quantity in rabbits
Hara S, et al.
Molecular Vision, 16(4), 2279-2279 (2010)
W W Schlaepfer et al.
The Journal of cell biology, 74(1), 241-250 (1977-07-01)
Localization of antisera to neurofilament antigens derived from rat peripheral nerve was carried out in tissues of rat and human peripheral and central nervous systems by indirect immunofluorescence. Unfixed and chloroform-methanol-fixed frozen sections of tissues were incubated in purified IgG
Ji-Young Bae et al.
Nutrition research and practice, 1(4), 279-284 (2007-01-01)
Solar ultraviolet (UV) irradiation leads to distinct changes in the skin connective tissues by degradation of collagen, which is a major structural component in the extracellular matrix. UV irradiation induces the production of matrix metalloproteinases (MMP) capable of attacking native
Migration, matrix production and lamellar bone formation of human osteoblast-like cells in porous titanium implants
Frosch KH, et al.
Cells Tissues Organs, 170(4), 214-227 (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service