Skip to Content
Merck

Skip To

F8052

Fructooligosaccharides from chicory

≥90% (as FOS and inulin)

Synonym(s):

FOS

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
12352201
NACRES:
NA.25
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

plant (Chicorium intybus)

Quality Level

assay

≥90% (as FOS and inulin)

form

powder

color

white

storage temp.

2-8°C

General description

Fructooligosaccharides are composed of glucose-(fructose)n with β−2→1 linkage between the fructose monomer units. The length of the fructose chain varies from 2 to 60 with an average degree of polymerization of >10. Fructooligosacchrides are resistant to hydrolysis by the intestinal enzymes, and have been shown to be indigestible by enzymes in the human small intestine but are fermented extensively by microflora in the lower digestive tract.

Application

Fructooligosaccharides from chicory may be used in prebiotic nutrition research to study the beneficial effects of fructooligosaccharides (FOS) on physiological processes and health. Fructooligosaccharide may be used in combinations with probiotic bacteria to assess their benefits as intestinal supplements.[1] Fructooligosaccharide may be used in mannose-binding lectin (MBL) research.[2]

Other Notes

Inulin has a greater chain length than fructooligosaccharides.
To gain a comprehensive understanding of our extensive range of Oligosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
I2255L852510115
biological source

plant (Chicorium intybus)

biological source

plant (chicory)

biological source

fungus (Candida rugosa)

biological source

-

assay

≥90% (as FOS and inulin)

assay

-

assay

-

assay

-

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

form

powder

form

powder

form

lyophilized powder

form

powder

storage temp.

2-8°C

storage temp.

room temp

storage temp.

2-8°C

storage temp.

2-8°C

color

white

color

white

color

-

color

-


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Mohd Anis Ganaie et al.
Applied biochemistry and biotechnology, 172(4), 2143-2159 (2013-12-18)
Fructooligosaccharides are influential prebiotics that affect various physiological functions in such a way that they promote positive impact to health. They occur naturally in many fruits and vegetables in trace amounts. However, they are mainly produced commercially by the reaction
Jiradej Manosroi et al.
Journal of food science and technology, 51(2), 341-346 (2014-02-05)
Fructooligosaccharide (FOS), a prebiotic was extracted from the grain of Coix lachryma-jobi Linn. (Job's tears) by hot water extraction at 60 °C for 1 h. The resulting dried powder extract was assayed for FOS content of 1-kestose (GF2), nystose (GF3) and 1-β-D-fructofuranosylnystose
Jacek Piatek et al.
International journal of environmental research and public health, 17(9) (2020-05-15)
A variety of activities potentially contribute to the beneficial effects of probiotic bacteria observed in humans. Among these is a direct inhibition of the growth of pathogenic bacteria in the gut. The present study characterizes head-to-head the in-vitro pathogen growth



Global Trade Item Number

SKUGTIN
F8052-50G04061833619995

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters