Skip to Content
Merck
All Photos(1)

Key Documents

10115

Sigma-Aldrich

Amyloglucosidase from Aspergillus niger

greener alternative

lyophilized, powder, ~70 U/mg

Synonym(s):

1,4-α-D-Glucan glucohydrolase, Exo-1,4-α-glucosidase, Glucoamylase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

form

powder

quality

lyophilized

specific activity

~70 U/mg

mol wt

Mr ~97,000

greener alternative product characteristics

Waste Prevention
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

parameter

temperature at 50 °C (optimum temperature)

greener alternative category

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency and waste prevention when used in starch hydrolysis research. For more information see the article in biofiles.

Amyloglucosidase is a disaccharide enzyme belonging to the α-glucosidase type. It is produced by several species of Aspergillus genus.

Application

Amyloglucosidase from Aspergillus niger has been used in glycogen assay and in acetate buffer for starch hydrolysis. It can also be used for glycogen determination in whole yeast cells and synthesis of hetero-oligosaccharides by glucoamylase in reverse.

Unit Definition

One unit corresponds to the amount of enzyme which liberates 1 μmole of glucose per minute at pH 4.8 and 60 °C (starch acc. to Zulkowsky, Cat. No. 85642, as substrate).

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sifang Liao et al.
Insect biochemistry and molecular biology, 133, 103495-103495 (2020-11-11)
Excess consumption of high-fat diet (HFD) is likely to result in obesity and increases the predisposition to associated health disorders. Drosophila melanogaster has emerged as an important model to study the effects of HFD on metabolism, gut function, behavior, and
Stem girdling manipulates leaf sugar concentrations and anthocyanin expression in sugar maple trees during autumn
Murakami PF, et al.
Tree physiology, 28(10), 1467-1473 (2008)
Trehalose is a key determinant of the quiescent metabolic state that fuels cell cycle progression upon return to growth
Shi Lei, et al.
Molecular Biology of the Cell, 21(12), 1982-1990 (2010)
Nicole M Grant et al.
Journal of experimental botany, 59(3), 705-714 (2008-02-07)
The relationships between heat production, alternative oxidase (AOX) pathway flux, AOX protein, and carbohydrates during floral development in Nelumbo nucifera (Gaertn.) were investigated. Three distinct physiological phases were identified: pre-thermogenic, thermogenic, and post-thermogenic. The shift to thermogenic activity was associated
Biodiversity: New Leads for the Pharmaceutical and Agrochemical Industries, 183-183 (2000)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service