Skip to Content
Merck

Skip To

F4759

Bovine Fibronectin

from bovine plasma, powder, suitable for cell culture

Fibronectin bovine plasma

Synonym(s):

Fibronectin

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

UNSPSC Code:
12352202
eCl@ss:
42020141
NACRES:
NA.75
EC Number:
289-149-2
MDL number:
Biological source:
bovine plasma
Form:
powder
Technique(s):
cell culture | mammalian: suitable
Concentration:
≥1 mg protein/mL
Mol wt:
~450 kDa
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Fibronectin bovine plasma, powder, BioReagent, suitable for cell culture

biological source

bovine plasma

Quality Segment

sterility

sterile

product line

BioReagent

form

powder

mol wt

~450 kDa

packaging

pkg of 1 mg, pkg of 2 mg, pkg of 5 mg

concentration

≥1 mg protein/mL

technique(s)

cell culture | mammalian: suitable

surface coverage

1‑5 μg/cm2

solubility

H2O: 1 mg/mL at 37 °C (Store reconstituted solution in working aliquots at -20°C or lower.)

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

bovine ... FN1(2807940)

General description

Fibronectin is a glycoprotein with multiple domains. It comprises a N-terminal, central binding domain and a heparin-binding domain. Bovine plasma fibronectin has polypeptide chains with disulfide linkage and corresponds to a molecular weight of 220 and 215 kDa.
Fibronectins are adhesive glycoproteins and are found both as cell surface proteins and in plasma. Fibronectin (FN) is a multifunctional glycoprotein, the gene of which is localized to human chromosome 2q34-36. This gene spans 75kb and contains 50 exons. It exists as two major isoforms, one soluble and one insoluble form. The former is present in plasma, whereas the insoluble form resides in tissues and extracellular matrix (ECM) of cartilage. There are around 20 different forms of FN due to alternative splicing of a variable region and two type III exons called Extra Domains A and B.[1]

Application

Fibronectin from bovine plasma is used for the following applications:
  • Used for coating the glass coverslips for the culture of decidual stromal cells (DEC)[2]
  • Cell movement assays (culture plates were coated with fibronectin)[3]
  • Cell spreading and immunofluorescence[4]
  • Protein specificity[5]
  • Integration of sensor films into microfluidic chips[6]
  • Histological and Immunohistochemical (IHC) analyses[7]
  • Used in primary cell cultures from patients with lung carcinoma[8]

Biochem/physiol Actions

Fibronectin is a regulator of protein scaffolding. It coordinates cellular processes like platelet functions, wound-healing, adhesion, migration and aggregation.Study shows that human dental pulp (HDP) cells rely on fibronectin (FN) for initial attachment and subsequent spreading. FN may exhibit mitogenic activity, promoting cell proliferation, and also play a role in regulating the differentiation process of pulp cells into odontoblasts. The presence of FN appears to influence the fate and development of HDP cells, specifically in their differentiation into odontoblasts, which are responsible for dentin formation in teeth.

Disclaimer

As supplied, this product should be stored at -20°C and will remain active for at least 2 years.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
F1141F2006F1056
technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

sterility

sterile

sterility

sterile-filtered

sterility

sterile

sterility

sterile

biological source

bovine plasma

biological source

bovine plasma

biological source

human plasma

biological source

human plasma

concentration

≥1 mg protein/mL

concentration

-

concentration

>1 mg/mL protein

concentration

-

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

mol wt

~450 kDa

mol wt

-

mol wt

450 kDa

mol wt

-


Still not finding the right product?

Explore all of our products under Fibronectin bovine plasma


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library







Global Trade Item Number

SKUGTIN
F4759-1MG04061833617847
F4759-5MG04061833617861
F4759-2MG04061833617854

Questions

Reviews

Active Filters

  1. charleston SC
    • Review 1
    • Vote 1
    5 out of 5 stars.

    culture reagent

    this works great for coating plates for cell culture

    Helpful?