Skip to Content
Merck
All Photos(1)

Key Documents

F1056

Sigma-Aldrich

Human Fibronectin

from human plasma, powder, suitable for cell culture

Synonym(s):

Fibronectin

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.75
Pricing and availability is not currently available.

Product Name

Fibronectin human plasma, lyophilized powder, BioReagent, suitable for cell culture, ≥95% (SDS-PAGE)

biological source

human plasma

Quality Level

sterility

sterile

product line

BioReagent

Assay

≥95% (SDS-PAGE)

form

lyophilized powder

packaging

pkg of 1 mg
pkg of 2 mg
pkg of 5 mg

technique(s)

cell culture | mammalian: suitable

surface coverage

1‑5 μg/cm2

solubility

H2O: 1 mg/mL at 37 °C

UniProt accession no.

Binding Specificity

Peptide Source: Collagen

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... FN1(2335)

Looking for similar products? Visit Product Comparison Guide

General description

Fibronectin is a class of high-molecular-weight adhesive glycoprotein,[1] predominant in extracellular matrix (ECM) and is widely distributed in tissue and blood. The FN gene, which is highly conserved, gives rise to a minimum of 20 isoforms through alternative splicing in humans. These isoforms include notable variants such as cellular FN and plasma FN.[2] Plasma FN is primarily synthesized in the liver, whereas cellular FN which represents only 5% of the circulating fibronectins[3] is locally produced by various cell types.[2]

Application

Fibronectin human plasma has been used:
  • in neural selection medium to coat tissue culture surface for neuronal differentiation[4]
  • to coat culture vessels for characterization of iPSC and iPSC‑derived BMEC in 2D cell culture[5]
  • to coat culture supports for endothelial cell culture[6]

Biochem/physiol Actions

Fibronectin plays a major role in biological processes. It interacts with surface receptors and other major ECM proteins like collagen to mediate cellular signals.[2] Fibronectin is involved in a variety of cellular functions, including adhesion, cytoskeletal organization, oncogenic transformation, cell migration, phagocytosis, hemostasis, and embryonic differentiation.[3] This glycoprotein can bind to integrins, collagens, fibrins and glycosaminoglycans (GAGs).[1] FN plays a crucial role in promoting cell cycle progression and proliferation. This interaction influences cyclins and immediate early response genes, contributing to cellular processes related to cell division and growth. FN is also involved in cell survival and protection from apoptosis in various cell types, including epithelial and endothelial cells and leukocytes. FN serves as a critical factor in promoting cell survival and preventing programmed cell death in these cellular contexts.[7] Mutations in the FN gene are associated with FN glomerulopathy and spondylometaphyseal dysplasia.[2] Fibronectin plays a significant role in the wound healing process, and in extracellular matrix (ECM) assembly, hence is considered suitable for tissue engineering applications.[1]

Caution

The reconstituted solution should be stored in working aliquots at -20°C or lower. Vortexing, excessive agitation, or additoinal freezing and thawing of reconstituted fibronectin are not recommended.

Preparation Note

This product is lyophilized from 0.05 M Tris buffered saline at pH 7.5. It is soluble at 1 mg/mL in water, and should be allowed to dissolve for at least 30 minutes at 37°C. A small amount of undissolved material may remain, but it will not affect performance. The protein homogeneity is evaluated by immunoelectrophoresis, is endotoxin tested, and the source material was tested negative for HIV, hepatitis B and hepatitis C.

In coating culture surfaces, fibronectin should be diluted in sterile balanced sterile salt solution and coated with minimal volume. Surface should then be air dried for 45 minutes at room temp and can be stored for 2-4 weeks at 2-8°C.

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fibronectin
Fabrizio M and Angela S
Encyclopedia of Immunology, 909-913 (1998)
Fibronectin
Breisnes HW, et al.
Biochemistry of Collagens, Laminins and Elastin (2024)
Tests to predict preeclampsia
Agustin CA, et al.
Journal of Separation Science (2015)
Vivek V Thacker et al.
eLife, 9 (2020-11-25)
We establish a murine lung-on-chip infection model and use time-lapse imaging to reveal the dynamics of host-Mycobacterium tuberculosis interactions at an air-liquid interface with a spatiotemporal resolution unattainable in animal models and to probe the direct role of pulmonary surfactant
Artificial biomaterials for urological tissue engineering
Farhat WA and Geutjes, PJ
Journal of Separation Science (2009)

Articles

Extracellular matrix proteins such as laminin, collagen, and fibronectin can be used as cell attachment substrates in cell culture.

Extracellular matrix proteins such as laminin, collagen, and fibronectin can be used as cell attachment substrates in cell culture.

Extracellular matrix proteins such as laminin, collagen, and fibronectin can be used as cell attachment substrates in cell culture.

Extracellular matrix proteins such as laminin, collagen, and fibronectin can be used as cell attachment substrates in cell culture.

Protocols

Dilute fibronectin for cell attachment, varying per cell type. Coating protocol, products, and FAQs provided.

Dilute fibronectin for cell attachment, varying per cell type. Coating protocol, products, and FAQs provided.

Dilute fibronectin for cell attachment, varying per cell type. Coating protocol, products, and FAQs provided.

Dilute fibronectin for cell attachment, varying per cell type. Coating protocol, products, and FAQs provided.

Questions

1–10 of 11 Questions  
  1. What is the molecular weight of this product?

    1 answer
    1. Human fibronectin is composed of two nearly identical disulfide bound peptides. Each has a molecular weight of 220 kDa for a combined total of 440 kDa.

      Helpful?

  2. This is to ask a question regarding coating flasks with the human freeze-dried fibronectin (F1056). After dissolving it in sterile water and diluting to the desired concentration should I incubate the flasks at 4 or 37 degrees overnight? Many thanks!

    1 answer
    1. As noted in the DESCRIPTION section of the Product Detail Page, fibronectin should be diluted in a sterile balanced sterile salt solution and coated with minimal volume. The surface should then be air dried for 45 minutes at room temp and can be stored for 2-4 weeks at 2-8°C. See the information below for a brief protocol:

      To coat culture surfaces:
      1. Dilute fibronectin in a sterile balanced salt solution
      and coat the culture surface (1-5 mg/cm2) with a
      minimal volume.
      2. Allow to air dry for at least 45 minutes at room
      temperature, Excess fibronectin may be removed
      by aspiration, but this is not necessary.
      3. Fibronectin coated cultureware can be stored for
      2-4 weeks at 2-8 °C in a closed sterile container or
      sterile sealable bags.

      Helpful?

  3. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  4. How long can I store the fibronectin solution?

    1 answer
    1. The product is good for at least a year at 2-8 °C if kept sterile.

      Helpful?

  5. How long can I store fibronectin coated plates, etc?

    1 answer
    1. Fibronectin coated cultureware can be stored for 2-4 weeks at 2-8 °C in a closed sterile container, or in sterile sealable bags.

      Helpful?

  6. How do I test the wells/flasks that have been coated with fibronectin?

    1 answer
    1. We test the coated culture surface by culturing cells.  We compare the adherence of the cells to wells that have been coated with media containing BSA (negative) and media containing 10% FBS (positive).  The cell growth should be not less than 50% of the positive control and greater than 50% of the negative control.

      Helpful?

  7. How do I solubilized Fibronectin?

    1 answer
    1. Dissolve at 1 mg/mL in water. Allow to dissolve for at least 30 minutes at 37 °C. A small amount of undissolved material may remain. This will not affect product performance.

      Helpful?

  8. What is the difference between Fibronectin Products F2006 and F1056?

    1 answer
    1. The difference between Product Nos. F2006 and F1056 is in the purity specifications and endotoxin testing.  Product  No. F2006 has a purity specification of ≥90% and is not tested for endotoxin contamination.  Product No. F1056 has a purity specification of ≥95% and is tested for endotoxin contamination with a specification of ≤100 EU/mg.

      Helpful?

  9. What is the procedure for coating culture surfaces with fibronectin?

    1 answer
    1. To coat culture surfaces: 1. Dilute fibronectin in sterile balanced salt solution and coat the culture surface (1-5 μg/cm2) with a minimal volume.  2. Allow to air dry for at least 45 minutes at room temperature.  Excess fibronectin may be removed by aspiration, but this is not necessary.

      Helpful?

  10. How do I determine how much fibronectin is in solution?

    1 answer
    1. As the product may contain undissolved material, the amount of fibronectin in solution can be determined by UV absorbance.  The extinction coefficient for a 1% solution (10 mg/ml) is 13.5 at 280 nm.

      Helpful?

1–10 of 11 Questions  

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service