Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C6H12O6
CAS Number:
Molecular Weight:
180.16
UNSPSC Code:
12352201
NACRES:
NA.21
PubChem Substance ID:
EC Number:
200-333-3
Beilstein/REAXYS Number:
1239004
MDL number:
biological source
corn
Quality Segment
product line
BioReagent
assay
≥99% (HPLC)
form
powder
optical activity
[α]25/D -91.3 to -87.7 °, c = 5% (w/v) in water
technique(s)
cell culture | insect: suitable, cell culture | mammalian: suitable
impurities
≤0.05% Glucose (enzymatic)
color
colorless
useful pH range
5.0-7.0 (25 °C, 18 g/L)
mp
119-122 °C (dec.) (lit.)
solubility
790 g/L at 20 °C
SMILES string
OC[C@@H](O)[C@@H](O)[C@H](O)C(=O)CO
InChI
1S/C6H12O6/c7-1-3(9)5(11)6(12)4(10)2-8/h3,5-9,11-12H,1-2H2/t3-,5-,6-/m1/s1
InChI key
BJHIKXHVCXFQLS-UYFOZJQFSA-N
General description
Fructose is a monosaccharide. It is present in fruits and vegetables. Fructose is the major carbohydrate in the diet. It binds with glucose to form sucrose. Excessive intake of fructose is associated with obesity, type 2 diabetes and cardiovascular disease.
Application
D-(-)-Fructose has been used for metabolite determination. It has also been used as a component of cultural media.
Other Notes
To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.
Still not finding the right product?
Explore all of our products under D-(−)-Fructose
Storage Class
11 - Combustible Solids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
Global Trade Item Number
| SKU | GTIN |
|---|---|
| F3510-500G | 04061835544882 |
| F3510-5KG | 04061833614006 |
| F3510-100G | 04061833613993 |