Skip to Content
Merck

Skip To

F0127

D-(−)-Fructose

≥99% (HPLC)

D-(−)-Fructose
1 of 1 reviewers received a sample product or took part in a promotion

Synonym(s):

D-Levulose, Fruit sugar

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C6H12O6
CAS Number:
Molecular Weight:
180.16
UNSPSC Code:
12352201
NACRES:
NA.21
PubChem Substance ID:
EC Number:
200-333-3
Beilstein/REAXYS Number:
1239004
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

corn

Quality Level

assay

≥99% (HPLC)

form

powder

technique(s)

HPLC: suitable

impurities

≤0.05% Glucose (enzymatic)

color

colorless

useful pH range

5.0-7 (25 °C, 18 g/L)

mp

119-122 °C (dec.) (lit.)

solubility

water: 790 g/L at 20 °C

storage temp.

room temp

SMILES string

OC[C@@H](O)[C@@H](O)[C@H](O)C(=O)CO

InChI

1S/C6H12O6/c7-1-3(9)5(11)6(12)4(10)2-8/h3,5-9,11-12H,1-2H2/t3-,5-,6-/m1/s1

InChI key

BJHIKXHVCXFQLS-UYFOZJQFSA-N

General description

D-fructose is a crucial ketohexose, which is commonly present in free form in fruit juices, musk, and honey accompanied by glucose. It is a constituent of raffinose, sucrose, and inulin. Fructose is formed by the hydrolysis of inulin.

Application

D-(-)-Fructose has been used:
  • as a supplement in modified Sabouraud (mSAB)/Sabouraud broth to culture Zygosaccharomyces bailii and to prepare ketchup to avoid the presence of any chemical preservatives
  • to prepare hypoosmotic fructose solution to assess the functional integrity of sperm membranes
  • to supplement drinking water to mimic high-fructose corn syrup containing soft drinks

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
F2543F279347748
biological source

corn

biological source

corn

biological source

corn

biological source

-

technique(s)

HPLC: suitable

technique(s)

HPLC: suitable

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

technique(s)

-

assay

≥99% (HPLC)

assay

≥99% (HPLC)

assay

≥99% (HPLC)

assay

-

Quality Level

300

Quality Level

200

Quality Level

100

Quality Level

100

solubility

water: 790 g/L at 20 °C

solubility

H2O: 1 M, clear, colorless

solubility

-

solubility

alcohol: soluble 1 gm in 15 ml, methanol: soluble 1 gm in 14 ml, H2O: freely soluble, acetone: freely soluble (Hot), acetone: slightly soluble, pyridine: soluble

form

powder

form

powder

form

-

form

solid


Still not finding the right product?

Explore all of our products under D-(−)-Fructose


Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



L Mertens et al.
Applied and environmental microbiology, 75(22), 7060-7069 (2009-09-29)
Within the field of predictive microbiology, the number of studies that quantify the effect of food structure on microbial behavior is very limited. This is mainly due to impracticalities related to the use of a nonliquid growth medium. In this
Anne C Rios et al.
Cancer cell, 35(4), 618-632 (2019-04-02)
Breast tumors are inherently heterogeneous, but the evolving cellular organization through neoplastic progression is poorly understood. Here we report a rapid, large-scale single-cell resolution 3D imaging protocol based on a one-step clearing agent that allows visualization of normal tissue architecture
Ariadna Montero-Blay et al.
Cell reports, 31(9), 107722-107722 (2020-06-04)
Here, we propose an approach to identify active metabolic pathways by integrating gene essentiality analysis and protein abundance. We use two bacterial species (Mycoplasma pneumoniae and Mycoplasma agalactiae) that share a high gene content similarity yet show significant metabolic differences.



Global Trade Item Number

SKUGTIN
F0127-1KG04061835366590
F0127-10MG04061835366583
F0127-100G04061835366576
F0127-5KG04061833610794
F0127-500G04061835366606

Questions

1–3 of 3 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. Is this producr RNase free?

    1 answer
    1. This product is not tested for RNase. Currently, an RNase/DNase free product is not available.

      Helpful?

  3. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

Active Filters

  1. 1 Ratings-only review